siRNA Details
Gene Symbol: GATSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145064.2 | NP_001138536.1 | 9 | 6772 | ||
GenBank mRNA | AK123110.1 | - | 1 | 3618 | ||
AL832384.1 | - | 2 | 3122 | |||
BC048422.1 | - | 1 | 1073 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
