siRNA Details
Gene Symbol: LOC105372751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135520.1 | 6 | 1693 | |||
GenBank mRNA | AK124194.1 | - | 6 | 1693 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
