siRNA Details
Gene Symbol: FKBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002014.3 | NP_002005.1 | 7 | 996 | ||
XM_011520929.2 | XP_011519231.1 | 7 | 1989 | |||
GenBank mRNA | AK313918.1 | - | 7 | 961 | ||
BC001786.1 | - | 7 | 951 | 3542330 | ||
BC002887.2 | - | 4 | 355 | |||
BC007924.2 | - | 7 | 896 | 4304462 | ||
M88279.1 | - | 7 | 897 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
