siRNA Details
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006985.2 | NP_008916.2 | 8 | 972 | ||
XM_011522748.2 | XP_011521050.1 | 8 | 1069 | |||
XM_011522749.2 | XP_011521051.1 | 9 | 1181 | |||
XM_011522750.2 | XP_011521052.1 | 7 | 987 | |||
XM_011522751.1 | XP_011521053.1 | 9 | 1138 | |||
XM_017023891.1 | XP_016879380.1 | 10 | 1251 | |||
GenBank mRNA | AK160377.1 | - | 28 | 5229 | ||
CR627265.1 | - | 8247 |
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277323.1 | NP_001264252.1 | 8 | 972 | ||
XM_005255484.3 | XP_005255541.1 | 11 | 1060 | |||
XM_011522576.2 | XP_011520878.2 | 10 | 3425 | |||
XM_011522577.2 | XP_011520879.2 | 10 | 3346 | |||
XM_011522578.2 | XP_011520880.2 | 8 | 1068 | |||
XM_011522580.2 | XP_011520882.2 | 8 | 999 | |||
XM_017023553.1 | XP_016879042.1 | 10 | 3435 | |||
XM_017023554.1 | XP_016879043.1 | 9 | 3348 |
Gene Symbol: NPIPB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306094.1 | NP_001293023.1 | 7 | 904 | ||
XM_011523496.2 | XP_011521798.1 | 8 | 960 | |||
XM_011523498.2 | XP_011521800.1 | 8 | 994 | |||
XM_011523501.2 | XP_011521803.1 | 4 | 786 | |||
XM_017023248.1 | XP_016878737.1 | 8 | 1397 | |||
XM_017023249.1 | XP_016878738.1 | 7 | 1214 |
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282524.1 | NP_001269453.1 | 7 | 989 | ||
XM_005255741.3 | XP_005255798.1 | 8 | 1801 | |||
XM_005255743.3 | XP_005255800.1 | 4 | 2380 | |||
XM_011545929.2 | XP_011544231.1 | 9 | 1621 | |||
XM_011545931.2 | XP_011544233.1 | 8 | 1254 | |||
XM_017023628.1 | XP_016879117.1 | 8 | 1004 | |||
XM_017023629.1 | XP_016879118.1 | 8 | 1547 | |||
XM_017023630.1 | XP_016879119.1 | 9 | 1274 | |||
XM_017023631.1 | XP_016879120.1 | 7 | 1618 | |||
XM_017023632.1 | XP_016879121.1 | 8 | 1004 | |||
GenBank mRNA | BC144439.1 | - | 7 | 950 | ||
BC157836.1 | - | 7 | 1639 | 9057055 | ||
KF175222.1 | - | 7 | 951 |
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310136.1 | NP_001297065.1 | 7 | 951 | ||
XM_017023622.1 | XP_016879111.1 | 8 | 1222 | |||
XM_017023623.1 | XP_016879112.1 | 8 | 1222 | |||
XM_017023624.1 | XP_016879113.1 | 8 | 1075 | |||
XM_017023625.1 | XP_016879114.1 | 8 | 1055 | |||
XM_017023626.1 | XP_016879115.1 | 9 | 1694 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287250.1 | NP_001274179.1 | 7 | 989 | ||
NM_001287251.1 | NP_001274180.1 | 8 | 1075 | |||
XM_003118719.7 | XP_003118767.2 | 8 | 1550 | |||
XM_011545690.2 | XP_011543992.1 | 10 | 1711 | |||
XM_011545693.2 | XP_011543995.1 | 9 | 1638 | |||
XM_017022814.1 | XP_016878303.1 | 9 | 1786 | |||
XM_017022815.1 | XP_016878304.1 | 9 | 1306 | |||
XM_017022816.1 | XP_016878305.1 | 9 | 1786 | |||
XM_017022817.1 | XP_016878306.1 | 8 | 1698 | |||
XM_017022818.1 | XP_016878307.1 | 8 | 3023 | |||
XM_017022819.1 | XP_016878308.1 | 9 | 1713 | |||
XM_017022820.1 | XP_016878309.1 | 9 | 1430 | |||
XM_017022821.1 | XP_016878310.1 | 8 | 2296 | |||
XM_017022822.1 | XP_016878311.1 | 9 | 3299 | |||
XM_017022823.1 | XP_016878312.1 | 7 | 2935 | |||
XM_017022824.1 | XP_016878313.1 | 9 | 1565 | |||
XM_017022825.1 | XP_016878314.1 | 9 | 1283 | |||
XM_017022826.1 | XP_016878315.1 | 8 | 1247 | |||
XM_017022827.1 | XP_016878316.1 | 4 | 994 | |||
GenBank mRNA | BC171825.1 | - | 8 | 1725 |
Gene Symbol: PDXDC2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647358.1 | - | 2 | 5552 |
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123721.1 | 29 | 5124 | |||
GenBank mRNA | AK131084.1 | - | 24 | 5488 | ||
AK131085.1 | - | 23 | 5326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
