siRNA Details
Gene Symbol: FAM74A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026802.1 | 1 | 386 | |||
NR_110998.1 | 1 | 386 | ||||
GenBank mRNA | AK127145.1 | - | 1 | 248 | ||
BC121813.1 | - | 1 | 240 |
Gene Symbol: FAM74A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026803.2 | 1 | 240 | |||
GenBank mRNA | BC118602.1 | - | 1 | 240 |
Gene Symbol: FAM74A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026801.1 | 1 | 420 |
Gene Symbol: FAM74A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110999.1 | 1 | 240 |
Gene Symbol: FAM74A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126166.1 | 1 | 420 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
