siRNA Details
Gene Symbol: PTCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545449.2 | XP_011543751.1 | 4 | 9539 | ||
GenBank mRNA | AK126657.1 | - | 1 | 453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
