siRNA Details
Gene Symbol: UBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033243.2 | 2 | 318 | |||
XM_017014802.1 | XP_016870291.1 | 1 | 76 | |||
GenBank mRNA | BP237024.1 | - | 2 | 286 | ||
BX473128.1 | - | 2 | 310 |
Gene Symbol: AREL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011537415.2 | XP_011535717.1 | 2 | 211 | ||
XM_011537417.2 | XP_011535719.1 | 2 | 220 | |||
GenBank mRNA | BX248747.1 | - | 2 | 225 |
Gene Symbol: ARFRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833084.2 | - | 1 | 3412 |
Gene Symbol: CCDC28A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011535728.2 | XP_011534030.1 | 4 | 1788 |
Gene Symbol: CLYBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011521051.2 | XP_011519353.1 | 9 | 1663 | ||
XM_017020402.1 | XP_016875891.1 | 8 | 1561 | |||
XM_017020404.1 | XP_016875893.1 | 8 | 2531 |
Gene Symbol: CYB561A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017345.1 | XP_016872834.1 | 2 | 1759 | ||
XM_017017346.1 | XP_016872835.1 | 2 | 1759 |
Gene Symbol: DCAF16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005248169.3 | XP_005248226.1 | 3 | 6686 | ||
XM_005248171.3 | XP_005248228.1 | 2 | 6582 | |||
XM_006713966.3 | XP_006714029.1 | 4 | 6857 | |||
XM_017008327.1 | XP_016863816.1 | 2 | 8151 | |||
XM_017008328.1 | XP_016863817.1 | 2 | 8136 |
Gene Symbol: DGKE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025243.1 | XP_016880732.1 | 5 | 1264 |
Gene Symbol: FAM208B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005252474.3 | XP_005252531.1 | 1 | 3524 | ||
XM_011519515.2 | XP_011517817.1 | 1 | 3524 | |||
XM_011519516.2 | XP_011517818.1 | 1 | 3524 | |||
XM_011519519.2 | XP_011517821.1 | 1 | 3524 | |||
XM_011519520.2 | XP_011517822.1 | 1 | 3524 | |||
XM_017016361.1 | XP_016871850.1 | 1 | 3524 | |||
GenBank mRNA | AL049233.1 | - | 1 | 1629 |
Gene Symbol: GPR78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080819.4 | NP_543009.2 | 3 | 3197 | ||
NR_045511.2 | 3 | 2504 |
Gene Symbol: LINC01588 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126496.1 | 1 | 1897 | |||
GenBank mRNA | AK090420.1 | - | 1 | 1897 |
Gene Symbol: LOC440600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036595.1 | 2 | 259 | |||
GenBank mRNA | BC053344.1 | - | 2 | 259 |
Gene Symbol: MAPK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002754.4 | NP_002745.1 | 12 | 2708 | ||
NR_072996.1 | 10 | 2556 |
Gene Symbol: MRPL30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145212.3 | NP_660213.1 | 6 | 1527 | ||
GenBank mRNA | BC022391.1 | - | 6 | 1492 | 4611185 |
Gene Symbol: PDCD4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026932.1 | 1 | 885 | |||
GenBank mRNA | AK093606.1 | - | 1 | 885 | ||
AK097703.1 | - | 1 | 1760 |
Gene Symbol: PGM5P3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121190.1 | 2 | 190 | |||
GenBank mRNA | CA406616.1 | - | 2 | 185 | ||
DA859889.1 | - | 2 | 130 |
Gene Symbol: PGM5P4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121185.1 | 2 | 207 | |||
NR_121186.1 | 2 | 207 | ||||
GenBank mRNA | CB266081.1 | - | 2 | 153 | ||
CF126671.1 | - | 2 | 174 | |||
CF127520.1 | - | 2 | 152 | |||
DA931978.1 | - | 2 | 207 |
Gene Symbol: RPP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104546.1 | NP_001098016.1 | 12 | 915 | ||
XM_011539120.2 | XP_011537422.1 | 15 | 1468 | |||
XM_011539121.2 | XP_011537423.1 | 15 | 1468 | |||
XM_017015475.1 | XP_016870964.1 | 15 | 1468 | |||
XM_017015476.1 | XP_016870965.1 | 14 | 1438 |
Gene Symbol: SEH1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX538037.1 | - | 1 | 1653 |
Gene Symbol: SLC17A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK075320.1 | - | 7 | 969 |
Gene Symbol: SLC35F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005263799.3 | XP_005263856.1 | 16 | 6904 | ||
XM_011511921.2 | XP_011510223.1 | 17 | 6355 | |||
XM_011511922.2 | XP_011510224.1 | 17 | 6572 | |||
XM_011511923.2 | XP_011510225.1 | 16 | 6257 | |||
XM_017005028.1 | XP_016860517.1 | 15 | 6218 |
Gene Symbol: USP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018052.1 | XP_016873541.1 | 12 | 4237 |
Gene Symbol: WLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537492.1 | - | 1 | 1570 |
Gene Symbol: YEATS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK001063.1 | - | 1 | 523 |
Gene Symbol: YPEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR933630.1 | - | 1 | 3147 |
Gene Symbol: ZADH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306093.1 | NP_001293022.1 | 2 | 6121 | ||
NM_175907.5 | NP_787103.1 | 2 | 6299 |
Gene Symbol: ZNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095796.1 | - | 3 | 513 |
Gene Symbol: ZNF317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL834126.2 | - | 5 | 1047 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
