siRNA Details
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK298381.1 | - | 5 | 908 | ||
BC039707.1 | - | 9 | 974 | 4479933 | ||
KF175210.1 | - | 9 | 937 | |||
KF175211.1 | - | 10 | 1011 | |||
KF175219.1 | - | 8 | 841 |
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522751.1 | XP_011521053.1 | 8 | 751 | ||
XM_017023891.1 | XP_016879380.1 | 9 | 864 | |||
GenBank mRNA | AK160377.1 | - | 27 | 4842 | ||
AK294177.1 | - | 3 | 628 | |||
KF175165.1 | - | 8 | 691 | |||
KF175166.1 | - | 7 | 583 | |||
KF175167.1 | - | 8 | 691 | |||
KF175169.1 | - | 8 | 655 | |||
KF175209.1 | - | 9 | 873 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255492.4 | XP_005255549.1 | 8 | 1059 | ||
XM_006720917.3 | XP_006720980.2 | 10 | 3118 | |||
XM_011522593.1 | XP_011520895.1 | 10 | 3118 | |||
XM_011522594.2 | XP_011520896.1 | 10 | 3118 | |||
XM_011522597.1 | XP_011520899.1 | 9 | 3010 | |||
GenBank mRNA | KF175213.1 | - | 7 | 348 |
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255485.4 | XP_005255542.1 | 8 | 1059 | ||
XM_011522581.1 | XP_011520883.1 | 10 | 3118 | |||
XM_011522583.2 | XP_011520885.1 | 10 | 3118 | |||
XM_011522586.1 | XP_011520888.1 | 9 | 3010 |
Gene Symbol: NPIPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522336.2 | XP_011520638.1 | 10 | 1059 | ||
GenBank mRNA | KF175168.1 | - | 8 | 652 | ||
KF175171.1 | - | 8 | 649 | |||
KF175172.1 | - | 8 | 655 |
Gene Symbol: NPIPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022837.1 | XP_016878326.1 | 9 | 728 | ||
XM_017022838.1 | XP_016878327.1 | 8 | 1123 | |||
XM_017022839.1 | XP_016878328.1 | 8 | 620 |
Gene Symbol: NPIPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720830.3 | XP_006720893.1 | 8 | 1675 | ||
XM_017022845.1 | XP_016878334.1 | 9 | 728 | |||
XM_017022846.1 | XP_016878335.1 | 8 | 620 |
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308629.1 | - | 1 | 880 |
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023627.1 | XP_016879116.1 | 8 | 1254 |
Gene Symbol: PDXDC2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003610.1 | 25 | 2988 | |||
GenBank mRNA | AL833251.1 | - | 25 | 2988 | ||
BC144350.1 | - | 6 | 1099 | 9052870 | ||
BX647358.1 | - | 2 | 4474 | |||
KF175220.1 | - | 7 | 509 |
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123721.1 | 27 | 4748 | |||
NR_123722.1 | 17 | 2809 | ||||
GenBank mRNA | AK131084.1 | - | 22 | 5112 | ||
AK131085.1 | - | 21 | 5018 | |||
AK302708.1 | - | 17 | 2809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
