siRNA Details
Gene Symbol: ZNF888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310127.1 | NP_001297056.1 | 2 | 158 | ||
XM_005259451.4 | XP_005259508.1 | 4 | 390 | |||
XM_017026798.1 | XP_016882287.1 | 4 | 551 | |||
XM_017026799.1 | XP_016882288.1 | 4 | 436 | |||
XM_017026800.1 | XP_016882289.1 | 2 | 167 |
Gene Symbol: ZNF320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207333.2 | NP_997216.2 | 3 | 291 | ||
XM_006723059.3 | XP_006723122.1 | 4 | 337 | |||
XM_017026393.1 | XP_016881882.1 | 5 | 582 | |||
XM_017026394.1 | XP_016881883.1 | 5 | 507 | |||
XM_017026395.1 | XP_016881884.1 | 5 | 655 | |||
XM_017026396.1 | XP_016881885.1 | 4 | 424 | |||
XM_017026397.1 | XP_016881886.1 | 4 | 429 | |||
XM_017026398.1 | XP_016881887.1 | 4 | 308 | |||
XM_017026399.1 | XP_016881888.1 | 3 | 301 | |||
XM_017026400.1 | XP_016881889.1 | 3 | 221 | |||
GenBank mRNA | AK307375.1 | - | 4 | 338 | ||
BC064408.1 | - | 4 | 518 | |||
BC105738.1 | - | 7 | 603 | |||
BC131555.1 | - | 2 | 146 | 40108031 | ||
CR933622.1 | - | 1 | 78 |
Gene Symbol: ZNF701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172655.1 | NP_001166126.1 | 4 | 523 | ||
NM_018260.2 | NP_060730.2 | 3 | 229 | |||
XM_011527092.1 | XP_011525394.1 | 3 | 303 | |||
GenBank mRNA | AK001753.1 | - | 3 | 227 | ||
BC054884.1 | - | 3 | 197 | |||
BC067346.1 | - | 4 | 523 | |||
BC080612.1 | - | 2 | 185 | 5816672 | ||
BC131719.1 | - | 3 | 203 | 40123413 | ||
BC136425.1 | - | 3 | 333 | 9020414 | ||
BC136426.1 | - | 3 | 201 | 9020415 |
Gene Symbol: ZNF83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DB272526.1 | - | 4 | 304 | ||
DT216882.1 | - | 6 | 507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
