siRNA Details
Gene Symbol: ZNF761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008401.3 | NP_001008401.3 | 4, 5 | 481 | ||
NM_001289951.1 | NP_001276880.1 | 3, 4 | 262 | |||
NM_001289952.1 | NP_001276881.1 | 3, 4 | 330 | |||
NM_001289953.1 | NP_001276882.1 | 2, 3 | 207 | |||
NR_110551.1 | 4, 5 | 385 | ||||
GenBank mRNA | AB107355.2 | - | 3, 4 | 331 | ||
BC071639.1 | - | 1, 2 | 55 | |||
BC103708.1 | - | 4, 5 | 366 | |||
DA237674.1 | - | 4, 5 | 481 | |||
DB232487.1 | - | 3, 4 | 254 |
Gene Symbol: ZNF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006969.3 | NP_008900.3 | 2, 3 | 180 | ||
NR_036599.1 | 2, 3 | 180 | ||||
XM_006723354.3 | XP_006723417.1 | 2, 3 | 197 | |||
XM_011527262.2 | XP_011525564.1 | 2, 3 | 210 | |||
GenBank mRNA | AK304704.1 | - | 2, 3 | 144 | ||
BC070146.1 | - | 2, 3 | 144 | |||
BX485233.1 | - | 2, 3 | 192 | |||
BX489697.1 | - | 2, 3 | 143 | |||
CN263784.1 | - | 2, 3 | 127 | |||
CR933598.1 | - | 2, 3 | 143 |
Gene Symbol: ZNF320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK307375.1 | - | 3, 4 | 261 |
Gene Symbol: ZNF468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008801.1 | NP_001008801.1 | 2, 3 | 174 | ||
NM_001277120.1 | NP_001264049.1 | 2, 3 | 174 | |||
GenBank mRNA | AK289841.1 | - | 2, 3 | 165 | ||
AK292449.1 | - | 2, 3 | 143 | |||
AY442330.1 | - | 2, 3 | 175 | |||
AY442331.1 | - | 2, 3 | 175 | |||
CR936599.1 | - | 2, 3 | 140 | |||
CR936695.1 | - | 2, 3 | 144 |
Gene Symbol: ZNF525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003699.1 | 2, 3 | 108 | |||
GenBank mRNA | AK097037.1 | - | 2, 3 | 128 | ||
DB221179.1 | - | 2, 3 | 108 |
Gene Symbol: ZNF765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040185.1 | NP_001035275.1 | 2, 3 | 146 | ||
GenBank mRNA | AK293890.1 | - | 2, 3 | 145 | ||
AK316300.1 | - | 2, 3 | 140 | |||
BC001610.1 | - | 1, 2 | 108 | |||
BC017357.1 | - | 1, 2 | 109 | |||
BC140724.1 | - | 2, 3 | 120 | 9021592 | ||
BX647981.1 | - | 2, 3 | 138 | |||
CD656338.1 | - | 2, 3 | 167 |
Gene Symbol: ZNF813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004301.3 | NP_001004301.2 | 2, 3 | 151 | ||
GenBank mRNA | DB235327.1 | - | 2, 3 | 142 |
Gene Symbol: ZNF83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105549.1 | NP_001099019.1 | 2, 3 | 176 | ||
NM_001105550.1 | NP_001099020.1 | 2, 3 | 176 | |||
NM_001105551.1 | NP_001099021.1 | 2, 3 | 176 | |||
NM_001105552.1 | NP_001099022.1 | 2, 3 | 176 | |||
NM_001277945.1 | NP_001264874.1 | 4, 5 | 444 | |||
NM_001277948.1 | NP_001264877.1 | 3, 4 | 317 | |||
NM_001277952.1 | NP_001264881.1 | 2, 3 | 176 | |||
XM_017026951.1 | XP_016882440.1 | 4, 5 | 422 | |||
GenBank mRNA | AK075066.1 | - | 4, 5 | 351 | ||
BC032867.1 | - | 2, 3 | 137 | 5264072 | ||
BX641726.1 | - | 2, 3 | 136 | |||
DA151844.1 | - | 2, 3 | 133 | |||
DA183557.1 | - | 2, 3 | 176 | |||
DA258790.1 | - | 2, 3 | 131 | |||
DA341245.1 | - | 2, 3 | 148 | |||
DA449506.1 | - | 2, 3 | 141 | |||
DB134270.1 | - | 3, 4 | 227 | |||
DB252431.1 | - | 2, 3 | 150 | |||
DB254966.1 | - | 4, 5 | 352 | |||
DB272526.1 | - | 3, 4 | 227 | |||
DT216882.1 | - | 5, 6 | 430 |
Gene Symbol: ZNF845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321522.1 | NP_001308451.1 | 3, 4 | 325 | ||
NM_001321523.1 | NP_001308452.1 | 3, 4 | 264 | |||
NM_001321524.1 | NP_001308453.1 | 3, 4 | 272 | |||
NM_138374.2 | NP_612383.1 | 2, 3 | 170 | |||
GenBank mRNA | AK295079.1 | - | 3, 4 | 244 | ||
DC400704.1 | - | 2, 3 | 141 |
Gene Symbol: ZNF888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310127.1 | NP_001297056.1 | 1, 2 | 81 | ||
XM_005259451.4 | XP_005259508.1 | 3, 4 | 313 | |||
XM_017026798.1 | XP_016882287.1 | 3, 4 | 474 | |||
XM_017026799.1 | XP_016882288.1 | 3, 4 | 359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
