siRNA Details
Gene Symbol: ZNF320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207333.2 | NP_997216.2 | 3 | 243 | ||
XM_006723059.3 | XP_006723122.1 | 4 | 289 | |||
XM_017026393.1 | XP_016881882.1 | 5 | 534 | |||
XM_017026394.1 | XP_016881883.1 | 5 | 459 | |||
XM_017026395.1 | XP_016881884.1 | 5 | 607 | |||
XM_017026396.1 | XP_016881885.1 | 4 | 376 | |||
XM_017026397.1 | XP_016881886.1 | 4 | 381 | |||
XM_017026398.1 | XP_016881887.1 | 4 | 260 | |||
XM_017026399.1 | XP_016881888.1 | 3 | 253 | |||
XM_017026400.1 | XP_016881889.1 | 3 | 173 | |||
GenBank mRNA | AK307375.1 | - | 4 | 290 | ||
BC064408.1 | - | 4 | 470 | |||
BC105738.1 | - | 7 | 555 | |||
BC131555.1 | - | 2 | 98 | 40108031 | ||
CR933622.1 | - | 1 | 30 |
Gene Symbol: ZNF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006969.3 | NP_008900.3 | 3 | 209 | ||
NR_036599.1 | 3 | 209 | ||||
XM_006723354.3 | XP_006723417.1 | 3 | 226 | |||
XM_011527262.2 | XP_011525564.1 | 3 | 239 | |||
GenBank mRNA | AK304704.1 | - | 3 | 173 | ||
BC070146.1 | - | 3 | 173 | |||
BX485233.1 | - | 3 | 221 | |||
BX489697.1 | - | 3 | 172 | |||
CN263784.1 | - | 3 | 156 | |||
CR933598.1 | - | 3 | 172 |
Gene Symbol: ZNF468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008801.1 | NP_001008801.1 | 3 | 203 | ||
NM_001277120.1 | NP_001264049.1 | 3 | 203 | |||
NR_102299.1 | 4 | 458 | ||||
NR_102300.1 | 4 | 439 | ||||
NR_102301.1 | 4 | 458 | ||||
XM_017027443.1 | XP_016882932.1 | 4 | 418 | |||
XM_017027444.1 | XP_016882933.1 | 4 | 418 | |||
GenBank mRNA | AK289841.1 | - | 3 | 194 | ||
AK292449.1 | - | 3 | 172 | |||
AY442330.1 | - | 3 | 204 | |||
AY442331.1 | - | 3 | 204 | |||
BX501408.1 | - | 4 | 410 | |||
CR936599.1 | - | 3 | 169 | |||
CR936695.1 | - | 3 | 173 | |||
DA245284.1 | - | 4 | 423 | |||
DB234532.1 | - | 4 | 429 |
Gene Symbol: ZNF525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003699.1 | 3 | 137 | |||
GenBank mRNA | AK097037.1 | - | 3 | 157 | ||
DB221179.1 | - | 3 | 137 |
Gene Symbol: ZNF534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143938.1 | NP_001137410.1 | 2 | 113 | ||
NM_001143939.1 | NP_001137411.1 | 2 | 113 | |||
NM_001291368.1 | NP_001278297.1 | 2 | 113 | |||
NM_001291369.1 | NP_001278298.1 | 2 | 113 | |||
GenBank mRNA | AB086839.1 | - | 2 | 113 | ||
AB086840.1 | - | 2 | 113 |
Gene Symbol: ZNF616 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178523.4 | NP_848618.2 | 3 | 332 | ||
NR_135823.1 | 3 | 332 | ||||
XM_017027438.1 | XP_016882927.1 | 1 | 2267 | |||
GenBank mRNA | AK092266.1 | - | 3 | 320 | ||
BC033199.1 | - | 3 | 311 | 4865984 | ||
BC125100.1 | - | 2 | 140 | |||
CD048470.1 | - | 5 | 459 | |||
DA735752.1 | - | 3 | 332 |
Gene Symbol: ZNF665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024733.3 | NP_079009.3 | 3 | 152 | ||
XM_005259266.4 | XP_005259323.1 | 3 | 186 | |||
XM_011527325.2 | XP_011525627.1 | 3 | 257 | |||
XM_011527327.2 | XP_011525629.1 | 4 | 561 | |||
GenBank mRNA | AK291403.1 | - | 3 | 142 | ||
AL832507.1 | - | 3 | 137 |
Gene Symbol: ZNF701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172655.1 | NP_001166126.1 | 4 | 475 | ||
NM_018260.2 | NP_060730.2 | 3 | 181 | |||
XM_011527092.1 | XP_011525394.1 | 3 | 255 | |||
GenBank mRNA | AK001753.1 | - | 3 | 179 | ||
BC054884.1 | - | 3 | 149 | |||
BC067346.1 | - | 4 | 475 | |||
BC080612.1 | - | 2 | 137 | 5816672 | ||
BC131719.1 | - | 3 | 155 | 40123413 | ||
BC136425.1 | - | 3 | 285 | 9020414 | ||
BC136426.1 | - | 3 | 153 | 9020415 |
Gene Symbol: ZNF738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027130.1 | 3 | 341 | |||
GenBank mRNA | AK027579.1 | - | 3 | 283 | ||
AK291002.1 | - | 3 | 280 | |||
AK291193.1 | - | 3 | 248 | |||
BC034499.1 | - | 3 | 349 |
Gene Symbol: ZNF761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008401.3 | NP_001008401.3 | 5 | 510 | ||
NM_001289951.1 | NP_001276880.1 | 4 | 291 | |||
NM_001289952.1 | NP_001276881.1 | 4 | 359 | |||
NM_001289953.1 | NP_001276882.1 | 3 | 236 | |||
NR_110551.1 | 5 | 414 | ||||
GenBank mRNA | AB107355.2 | - | 4 | 360 | ||
BC071639.1 | - | 2 | 84 | |||
BC103708.1 | - | 5 | 395 | |||
DA237674.1 | - | 5 | 510 | |||
DB232487.1 | - | 4 | 283 |
Gene Symbol: ZNF765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040185.1 | NP_001035275.1 | 3 | 175 | ||
GenBank mRNA | AK293890.1 | - | 3 | 174 | ||
AK316300.1 | - | 3 | 169 | |||
BC001610.1 | - | 2 | 137 | |||
BC017357.1 | - | 2 | 138 | |||
BC140724.1 | - | 3 | 149 | 9021592 | ||
BX647981.1 | - | 3 | 167 | |||
CD656338.1 | - | 3 | 196 |
Gene Symbol: ZNF766 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010851.2 | NP_001010851.1 | 2 | 98 | ||
XM_011527460.1 | XP_011525762.1 | 2 | 425 | |||
XM_017027440.1 | XP_016882929.1 | 2 | 293 | |||
GenBank mRNA | AK024074.1 | - | 2 | 76 | ||
AK097729.1 | - | 3 | 479 | |||
BC109201.1 | - | 2 | 223 | |||
BC136833.1 | - | 2 | 86 | 9020824 | ||
BC136834.1 | - | 2 | 86 | 9020825 | ||
BX538183.1 | - | 2 | 67 | |||
BX648899.1 | - | 2 | 99 |
Gene Symbol: ZNF813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004301.3 | NP_001004301.2 | 3 | 180 | ||
GenBank mRNA | DB235327.1 | - | 3 | 171 |
Gene Symbol: ZNF83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105549.1 | NP_001099019.1 | 3 | 205 | ||
NM_001105550.1 | NP_001099020.1 | 3 | 205 | |||
NM_001105551.1 | NP_001099021.1 | 3 | 205 | |||
NM_001105552.1 | NP_001099022.1 | 3 | 205 | |||
NM_001277945.1 | NP_001264874.1 | 5 | 473 | |||
NM_001277948.1 | NP_001264877.1 | 4 | 346 | |||
NM_001277949.1 | NP_001264878.1 | 4 | 385 | |||
NM_001277952.1 | NP_001264881.1 | 3 | 205 | |||
XM_017026951.1 | XP_016882440.1 | 5 | 451 | |||
GenBank mRNA | AK075066.1 | - | 5 | 380 | ||
BC032867.1 | - | 3 | 166 | 5264072 | ||
BX641726.1 | - | 3 | 165 | |||
DA151844.1 | - | 3 | 162 | |||
DA183557.1 | - | 3 | 205 | |||
DA258790.1 | - | 3 | 160 | |||
DA341245.1 | - | 3 | 177 | |||
DA434207.1 | - | 4 | 290 | |||
DA449506.1 | - | 3 | 170 | |||
DB134270.1 | - | 4 | 256 | |||
DB252431.1 | - | 3 | 179 | |||
DB254966.1 | - | 5 | 381 | |||
DB272526.1 | - | 4 | 256 | |||
DT216882.1 | - | 6 | 459 |
Gene Symbol: ZNF836 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102657.1 | NP_001096127.1 | 4 | 574 | ||
XM_011526558.2 | XP_011524860.1 | 4 | 416 | |||
XM_011526559.2 | XP_011524861.1 | 3 | 461 | |||
XM_017026389.1 | XP_016881878.1 | 4 | 349 | |||
GenBank mRNA | AK131308.1 | - | 4 | 426 | ||
BC011784.2 | - | 4 | 574 | |||
BC049172.1 | - | 4 | 423 |
Gene Symbol: ZNF888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310127.1 | NP_001297056.1 | 2 | 110 | ||
XM_005259451.4 | XP_005259508.1 | 4 | 342 | |||
XM_017026798.1 | XP_016882287.1 | 4 | 503 | |||
XM_017026799.1 | XP_016882288.1 | 4 | 388 | |||
XM_017026800.1 | XP_016882289.1 | 2 | 119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
