siRNA Details
Gene Symbol: ZNF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006969.3 | NP_008900.3 | 3 | 233 | ||
NR_036599.1 | 3 | 233 | ||||
XM_006723354.3 | XP_006723417.1 | 3 | 250 | |||
XM_011527262.2 | XP_011525564.1 | 3 | 263 | |||
GenBank mRNA | AK304704.1 | - | 3 | 197 | ||
BC070146.1 | - | 3 | 197 | |||
BC140720.1 | - | 1 | 16 | 9021588 | ||
BX485233.1 | - | 3 | 245 | |||
BX489697.1 | - | 3 | 196 | |||
CN263784.1 | - | 3 | 180 | |||
CR933598.1 | - | 3 | 196 |
Gene Symbol: ZNF320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207333.2 | NP_997216.2 | 3 | 267 | ||
XM_006723059.3 | XP_006723122.1 | 4 | 313 | |||
XM_017026393.1 | XP_016881882.1 | 5 | 558 | |||
XM_017026394.1 | XP_016881883.1 | 5 | 483 | |||
XM_017026395.1 | XP_016881884.1 | 5 | 631 | |||
XM_017026396.1 | XP_016881885.1 | 4 | 400 | |||
XM_017026397.1 | XP_016881886.1 | 4 | 405 | |||
XM_017026398.1 | XP_016881887.1 | 4 | 284 | |||
XM_017026399.1 | XP_016881888.1 | 3 | 277 | |||
XM_017026400.1 | XP_016881889.1 | 3 | 197 | |||
GenBank mRNA | AK307375.1 | - | 4 | 314 | ||
BC064408.1 | - | 4 | 494 | |||
BC105738.1 | - | 7 | 579 | |||
BC131555.1 | - | 2 | 122 | 40108031 | ||
CR933622.1 | - | 1 | 54 |
Gene Symbol: ZNF468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008801.1 | NP_001008801.1 | 3 | 227 | ||
NM_001277120.1 | NP_001264049.1 | 3 | 227 | |||
NR_102299.1 | 4 | 482 | ||||
NR_102300.1 | 4 | 463 | ||||
NR_102301.1 | 4 | 482 | ||||
XM_017027443.1 | XP_016882932.1 | 4 | 442 | |||
XM_017027444.1 | XP_016882933.1 | 4 | 442 | |||
GenBank mRNA | AK289841.1 | - | 3 | 218 | ||
AK292449.1 | - | 3 | 196 | |||
AY442330.1 | - | 3 | 228 | |||
AY442331.1 | - | 3 | 228 | |||
BX501408.1 | - | 4 | 434 | |||
CR936599.1 | - | 3 | 193 | |||
CR936695.1 | - | 3 | 197 | |||
DA245284.1 | - | 4 | 447 | |||
DB234532.1 | - | 4 | 453 |
Gene Symbol: ZNF525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003699.1 | 3 | 161 | |||
GenBank mRNA | AK097037.1 | - | 3 | 181 | ||
DB221179.1 | - | 3 | 161 |
Gene Symbol: ZNF701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172655.1 | NP_001166126.1 | 4 | 499 | ||
NM_018260.2 | NP_060730.2 | 3 | 205 | |||
XM_011527092.1 | XP_011525394.1 | 3 | 279 | |||
GenBank mRNA | AK001753.1 | - | 3 | 203 | ||
BC054884.1 | - | 3 | 173 | |||
BC067346.1 | - | 4 | 499 | |||
BC080612.1 | - | 2 | 161 | 5816672 | ||
BC131719.1 | - | 3 | 179 | 40123413 | ||
BC136425.1 | - | 3 | 309 | 9020414 | ||
BC136426.1 | - | 3 | 177 | 9020415 |
Gene Symbol: ZNF761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008401.3 | NP_001008401.3 | 5 | 534 | ||
NM_001289951.1 | NP_001276880.1 | 4 | 315 | |||
NM_001289952.1 | NP_001276881.1 | 4 | 383 | |||
NM_001289953.1 | NP_001276882.1 | 3 | 260 | |||
NR_110551.1 | 5 | 438 | ||||
GenBank mRNA | AB107355.2 | - | 4 | 384 | ||
BC071639.1 | - | 2 | 108 | |||
BC103708.1 | - | 5 | 419 | |||
DA237674.1 | - | 5 | 534 | |||
DB232487.1 | - | 4 | 307 |
Gene Symbol: ZNF765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040185.1 | NP_001035275.1 | 3 | 199 | ||
GenBank mRNA | AK293890.1 | - | 3 | 198 | ||
AK316300.1 | - | 3 | 193 | |||
BC001610.1 | - | 2 | 161 | |||
BC017357.1 | - | 2 | 162 | |||
BC140724.1 | - | 3 | 173 | 9021592 | ||
BX647981.1 | - | 3 | 191 | |||
CD656338.1 | - | 3 | 220 |
Gene Symbol: ZNF813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004301.3 | NP_001004301.2 | 3 | 204 | ||
GenBank mRNA | DB235327.1 | - | 3 | 195 |
Gene Symbol: ZNF83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105549.1 | NP_001099019.1 | 3 | 229 | ||
NM_001105550.1 | NP_001099020.1 | 3 | 229 | |||
NM_001105551.1 | NP_001099021.1 | 3 | 229 | |||
NM_001105552.1 | NP_001099022.1 | 3 | 229 | |||
NM_001277945.1 | NP_001264874.1 | 5 | 497 | |||
NM_001277948.1 | NP_001264877.1 | 4 | 370 | |||
NM_001277949.1 | NP_001264878.1 | 4 | 409 | |||
NM_001277952.1 | NP_001264881.1 | 3 | 229 | |||
XM_017026951.1 | XP_016882440.1 | 5 | 475 | |||
GenBank mRNA | AK075066.1 | - | 5 | 404 | ||
BC032867.1 | - | 3 | 190 | 5264072 | ||
BX641726.1 | - | 3 | 189 | |||
DA151844.1 | - | 3 | 186 | |||
DA183557.1 | - | 3 | 229 | |||
DA258790.1 | - | 3 | 184 | |||
DA341245.1 | - | 3 | 201 | |||
DA434207.1 | - | 4 | 314 | |||
DA449506.1 | - | 3 | 194 | |||
DA497861.1 | - | 3 | 253 | |||
DB134270.1 | - | 4 | 280 | |||
DB252431.1 | - | 3 | 203 | |||
DB254966.1 | - | 5 | 405 | |||
DB272526.1 | - | 4 | 280 | |||
DT216882.1 | - | 6 | 483 |
Gene Symbol: ZNF888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310127.1 | NP_001297056.1 | 2 | 134 | ||
XM_005259451.4 | XP_005259508.1 | 4 | 366 | |||
XM_017026798.1 | XP_016882287.1 | 4 | 527 | |||
XM_017026799.1 | XP_016882288.1 | 4 | 412 | |||
XM_017026800.1 | XP_016882289.1 | 2 | 143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
