siRNA Details
Gene Symbol: LINC00508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126452.2 | 4 | 497 | |||
GenBank mRNA | BC032019.1 | - | 1 | 186 | ||
DA807329.1 | - | 4 | 499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
