siRNA Details
Gene Symbol: ZNF397 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006722558.2 | XP_006722621.1 | 4 | 3337 | ||
XM_011526229.2 | XP_011524531.1 | 5 | 3426 | |||
XM_011526230.2 | XP_011524532.1 | 4 | 3628 | |||
XM_011526231.2 | XP_011524533.1 | 4 | 3225 | |||
XM_017026042.1 | XP_016881531.1 | 4 | 3280 |
Gene Symbol: ATR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB208847.1 | - | 9 | 1895 |
Gene Symbol: COQ7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190983.1 | NP_001177912.1 | 6 | 1129 | ||
NM_016138.4 | NP_057222.2 | 6 | 1180 | |||
GenBank mRNA | AK024291.1 | - | 6 | 1161 | ||
AK225115.1 | - | 6 | 1129 | |||
BC043272.1 | - | 6 | 1929 | 5297074 |
Gene Symbol: LINC00508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126452.2 | 4 | 449 | |||
GenBank mRNA | BC032019.1 | - | 1 | 138 | ||
DA807329.1 | - | 4 | 451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
