siRNA Details
Gene Symbol: IGKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF113887.1 | - | 2 | 114 | ||
BC034141.1 | - | 2 | 139 | |||
BC067092.1 | - | 2 | 131 | |||
BC070334.1 | - | 2 | 139 | |||
BC070335.1 | - | 2 | 140 | |||
BC070336.1 | - | 2 | 143 | |||
BC070361.1 | - | 2 | 143 | |||
BC072669.1 | - | 2 | 123 | |||
BC073764.1 | - | 2 | 125 | |||
BC073791.1 | - | 2 | 130 | |||
BC095489.1 | - | 2 | 134 | |||
BC110292.1 | - | 2 | 122 |
Gene Symbol: IGKV1D-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U86790.1 | - | 2 | 114 |
Gene Symbol: IGKV@ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF103383.1 | - | 1 | 37 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
