siRNA Details
Gene Symbol: POTEE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083538.1 | NP_001077007.1 | 2 | 615 | ||
XM_011511216.2 | XP_011509518.1 | 2 | 693 | |||
XM_017004159.1 | XP_016859648.1 | 2 | 694 | |||
XM_017004161.1 | XP_016859650.1 | 4 | 1052 | |||
GenBank mRNA | AY462868.1 | - | 2 | 615 | ||
AY462871.1 | - | 2 | 615 | |||
AY462872.1 | - | 2 | 615 |
Gene Symbol: POTEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099771.2 | NP_001093241.1 | 4 | 963 | ||
XM_017004833.1 | XP_016860322.1 | 2 | 708 |
Gene Symbol: POTEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005356.2 | NP_001005356.1 | 2 | 615 | ||
NR_027480.1 | 2 | 615 | ||||
GenBank mRNA | AY462874.1 | - | 2 | 615 | ||
AY465171.1 | - | 3 | 615 | |||
AY465172.1 | - | 3 | 615 | |||
BC127623.1 | - | 2 | 595 |
Gene Symbol: POTEH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136213.1 | NP_001129685.1 | 2 | 726 | ||
GenBank mRNA | AY466021.1 | - | 2 | 726 |
Gene Symbol: POTEKP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033885.2 | 1 | 54 | |||
GenBank mRNA | BM763453.1 | - | 1 | 54 |
Gene Symbol: POTEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145442.1 | NP_001138914.1 | 2 | 615 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
