siRNA Details
Gene Symbol: AP1S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039569.1 | NP_001034658.1 | 5 | 3178 | ||
NR_110905.1 | 6 | 3217 | ||||
NR_110906.1 | 4 | 3069 | ||||
XM_011510600.2 | XP_011508902.1 | 4 | 3020 |
Gene Symbol: FAM221A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127774.1 | - | 3 | 992 |
Gene Symbol: NME6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ020663.1 | - | 1 | 453 |
Gene Symbol: PARP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309863.1 | - | 3 | 1792 |
Gene Symbol: PDE3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209326.1 | - | 2 | 4596 |
Gene Symbol: S100PBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256121.1 | NP_001243050.1 | 7 | 2978 | ||
NM_022753.3 | NP_073590.2 | 7 | 2992 | |||
NR_135106.1 | 8 | 2891 | ||||
NR_135107.1 | 7 | 2848 | ||||
XM_011541961.2 | XP_011540263.1 | 7 | 2927 | |||
XM_011541962.2 | XP_011540264.1 | 7 | 3036 | |||
XM_017002098.1 | XP_016857587.1 | 7 | 2989 | |||
GenBank mRNA | AK022965.1 | - | 6 | 2326 | ||
AK025811.1 | - | 1 | 678 | |||
AK090711.1 | - | 7 | 2978 | |||
BX647916.1 | - | 7 | 2955 |
Gene Symbol: TBC1D7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318806.1 | NP_001305735.1 | 3 | 2068 | ||
GenBank mRNA | AL833474.1 | - | 3 | 2041 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
