siRNA Details
Gene Symbol: RUSC1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093295.1 | - | 4 | 1057 | ||
AK125510.1 | - | 1 | 2688 | |||
BC131614.1 | - | 2 | 1147 | 40115930 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
