siRNA Details
Gene Symbol: PTMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099285.1 | NP_001092755.1 | 1, 2 | 223 | ||
NM_002823.4 | NP_002814.3 | 1, 2 | 223 | |||
GenBank mRNA | AF348514.1 | - | 1, 2 | 41 | ||
AK223605.1 | - | 1, 2 | 218 | |||
AK314106.1 | - | 1, 2 | 218 | |||
AY169282.1 | - | 1, 2 | 41 | |||
BC003510.1 | - | 1, 2 | 206 | 3610808 | ||
BC022433.1 | - | 1, 2 | 222 | 4277490 | ||
BC034921.1 | - | 1, 2 | 214 | 4335961 | ||
BC051265.1 | - | 1, 2 | 206 | 3832454 | ||
BC066905.1 | - | 1, 2 | 144 | 4830132 | ||
BC070480.1 | - | 1, 2 | 208 | 6205582 | ||
BC071647.1 | - | 1, 2 | 201 | 5112610 | ||
BC071879.1 | - | 1, 2 | 203 | 6425103 | ||
BC098420.1 | - | 1, 2 | 208 | 5017164 | ||
BM839980.1 | - | 1, 2 | 223 | |||
M14483.1 | - | 1, 2 | 173 | |||
M14630.1 | - | 1, 2 | 218 | |||
M14794.1 | - | 1, 2 | 109 | |||
M26708.1 | - | 1, 2 | 196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
