siRNA Details
Gene Symbol: OPRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000914.4 | NP_000905.3 | 4 | 13362 | ||
NM_001145279.3 | NP_001138751.1 | 6 | 13721 | |||
NM_001145280.3 | NP_001138752.1 | 4 | 13152 | |||
NM_001145281.2 | NP_001138753.1 | 4 | 12693 | |||
NM_001145287.2 | NP_001138759.1 | 4 | 13927 | |||
NM_001285522.1 | NP_001272451.1 | 2 | 12488 | |||
NM_001285524.1 | NP_001272453.1 | 5 | 13257 | |||
NM_001285526.1 | NP_001272455.1 | 4 | 13908 | |||
NR_104348.1 | 5 | 13162 | ||||
NR_104350.1 | 3 | 12700 | ||||
XM_017010906.1 | XP_016866395.1 | 4 | 14837 |
Gene Symbol: A1BG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056201.1 | - | 2 | 990 | ||
EU794585.1 | - | 8 | 2242 |
Gene Symbol: ACAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127051.1 | - | 6 | 3684 |
Gene Symbol: APOBEC3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC061914.1 | - | 2 | 919 |
Gene Symbol: ARHGAP45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258328.2 | NP_001245257.1 | 23 | 4681 | ||
NM_001282334.1 | NP_001269263.1 | 13 | 4568 | |||
NM_001282335.1 | NP_001269264.1 | 22 | 4247 | |||
NM_001321232.1 | NP_001308161.1 | 23 | 4617 | |||
NM_012292.4 | NP_036424.2 | 23 | 4770 |
Gene Symbol: BRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017019995.1 | XP_016875484.1 | 1 | 18 |
Gene Symbol: CCAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU617955.1 | - | 1 | 575 |
Gene Symbol: CENPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006714064.2 | XP_006714127.1 | 19 | 5340 |
Gene Symbol: CFAP97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024878.1 | - | 1 | 1328 |
Gene Symbol: CRHR1-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026905.2 | 1 | 149 | |||
NR_026906.2 | 1 | 149 |
Gene Symbol: DLG2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF525782.1 | - | LIT2116 PSZA11q14 |
1 | 2084 |
Gene Symbol: EEF1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032378.4 | NP_115754.3 | 2 | 414 | ||
XM_006716523.1 | XP_006716586.1 | 2 | 237 | |||
XM_011516905.2 | XP_011515207.1 | 2 | 4859 | |||
XM_011516906.2 | XP_011515208.1 | 4 | 3451 | |||
GenBank mRNA | AK024550.1 | - | 2 | 152 | ||
AK225040.1 | - | 2 | 152 | |||
DA977213.1 | - | 2 | 152 |
Gene Symbol: EIF3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309944.1 | - | 5 | 694 | ||
AK311624.1 | - | 5 | 831 | |||
AK315896.1 | - | 5 | 700 |
Gene Symbol: FAM134C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178126.3 | NP_835227.1 | 9 | 2537 | ||
NR_026697.1 | 8 | 2506 | ||||
XM_011524440.2 | XP_011522742.1 | 9 | 2284 | |||
GenBank mRNA | AK057409.1 | - | 1 | 376 | ||
AK091125.1 | - | 8 | 2881 | |||
AK225797.1 | - | 9 | 2484 | |||
BC024980.1 | - | 1 | 369 | |||
BC049370.1 | - | 9 | 2494 | 5263052 | ||
BX647175.1 | - | 9 | 2428 | |||
BX648069.1 | - | 1 | 1478 |
Gene Symbol: FOXL2NB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040061.2 | NP_001035150.1 | 3 | 2885 | ||
XM_005247443.3 | XP_005247500.1 | 4 | 2971 | |||
GenBank mRNA | AK125319.1 | - | 3 | 2885 |
Gene Symbol: HNRNPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126950.1 | - | 1 | 740 |
Gene Symbol: JPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023484.1 | XP_016878973.1 | 2 | 2227 |
Gene Symbol: LINC01269 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125769.1 | 3, 4 | 450 | |||
GenBank mRNA | DA437066.1 | - | 3, 4 | 452 |
Gene Symbol: LINC01531 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040046.1 | 2 | 817 | |||
GenBank mRNA | AK128099.1 | - | 2 | 817 |
Gene Symbol: LMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127915.1 | - | 1 | 45 |
Gene Symbol: LOC100422781 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096219.1 | - | 2 | 1403 |
Gene Symbol: LOC101927040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055332.1 | - | 6 | 1026 |
Gene Symbol: LOC101928855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110852.1 | 4 | 2502 | |||
GenBank mRNA | AK123620.1 | - | 4 | 2502 |
Gene Symbol: LOC107985184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003082.1 | XP_016858571.1 | 2 | 2911 |
Gene Symbol: LOC729652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135198.1 | 1 | 1920 | |||
GenBank mRNA | AL834536.1 | - | 1 | 1920 |
Gene Symbol: LOC93429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127522.1 | - | 1 | 909 |
Gene Symbol: LUC7L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094081.1 | - | 1 | 1186 |
Gene Symbol: MAP7D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173516.1 | NP_001166987.1 | 19 | 3464 | ||
NM_024597.3 | NP_078873.2 | 19 | 3576 | |||
XM_005262472.1 | XP_005262529.1 | 19 | 3428 | |||
GenBank mRNA | AK022711.1 | - | 7 | 1257 |
Gene Symbol: MYLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096893.1 | - | 5 | 1282 |
Gene Symbol: NEAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131012.1 | 1 | 18637 | |||
GenBank mRNA | GQ859162.1 | - | 1 | 18637 |
Gene Symbol: OR51E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030774.3 | NP_110401.1 | 2 | 1856 | ||
GenBank mRNA | AY033942.1 | - | 2 | 1852 |
Gene Symbol: PARP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011512928.2 | XP_011511230.1 | 16 | 7213 |
Gene Symbol: PAX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936798.1 | - | 1 | 1242 |
Gene Symbol: RAB11FIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090437.1 | - | 2 | 4344 |
Gene Symbol: RNF138P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001575.1 | 1 | 1411 | |||
GenBank mRNA | BC040295.1 | - | 1 | 1410 |
Gene Symbol: RNF144B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182757.3 | NP_877434.2 | 8 | 4072 | ||
XM_005248986.3 | XP_005249043.1 | 7 | 3988 | |||
GenBank mRNA | AB076367.1 | - | 8 | 4057 | ||
AJ420555.1 | - | 1 | 737 | |||
BC063311.1 | - | 8 | 3944 | 30331343 |
Gene Symbol: RRAGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092808.1 | - | 1 | 280 |
Gene Symbol: RTBDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270440.1 | NP_001257369.1 | 7 | 1113 | ||
GenBank mRNA | AK093672.1 | - | 7 | 1113 |
Gene Symbol: TIMM23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290117.1 | NP_001277046.1 | 7 | 1734 | ||
NM_001290118.1 | NP_001277047.1 | 5 | 1590 | |||
NR_110767.1 | 8 | 1858 | ||||
GenBank mRNA | AK098044.1 | - | 8 | 1843 | ||
BU928648.1 | - | 1 | 129 |
Gene Symbol: TRIM69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK226115.1 | - | 1 | 470 |
Gene Symbol: TXNDC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024715.3 | NP_078991.3 | 5 | 3113 |
Gene Symbol: VHLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004319.2 | NP_001004319.1 | 1 | 74 | ||
GenBank mRNA | AA324790.1 | - | 1 | 74 |
Gene Symbol: ZCCHC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131766.1 | 2 | 195 | |||
GenBank mRNA | DA466245.1 | - | 2 | 136 |
Gene Symbol: ZNF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537507.1 | - | 1 | 2182 |
Gene Symbol: ZNF283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005258784.3 | XP_005258841.1 | 7 | 4476 | ||
XM_005258785.3 | XP_005258842.1 | 6 | 4270 | |||
XM_005258786.3 | XP_005258843.1 | 7 | 4435 | |||
XM_017026628.1 | XP_016882117.1 | 8 | 4557 | |||
XM_017026629.1 | XP_016882118.1 | 8 | 4598 | |||
XM_017026630.1 | XP_016882119.1 | 8 | 4561 | |||
XM_017026631.1 | XP_016882120.1 | 8 | 4428 | |||
XM_017026632.1 | XP_016882121.1 | 8 | 4604 | |||
XM_017026633.1 | XP_016882122.1 | 8 | 4553 | |||
XM_017026634.1 | XP_016882123.1 | 7 | 4306 | |||
XM_017026635.1 | XP_016882124.1 | 6 | 4371 | |||
XM_017026636.1 | XP_016882125.1 | 2 | 10076 | |||
XM_017026637.1 | XP_016882126.1 | 6 | 4349 | |||
XM_017026638.1 | XP_016882127.1 | 5 | 4143 | |||
XM_017026639.1 | XP_016882128.1 | 7 | 4471 |
Gene Symbol: ZNF554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK027860.1 | - | 1 | 1334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
