siRNA Details
Gene Symbol: LZIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316973.1 | NP_001303902.1 | 8 | 2517 | ||
NM_001316974.1 | NP_001303903.1 | 8 | 2460 | |||
NM_001316976.1 | NP_001303905.1 | 6 | 2339 | |||
NM_032368.4 | NP_115744.2 | 8 | 2617 | |||
XM_005263509.2 | XP_005263566.1 | 6 | 2334 | |||
XM_017002546.1 | XP_016858035.1 | 8 | 2363 | |||
XM_017002547.1 | XP_016858036.1 | 8 | 2383 |
Gene Symbol: BTD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006713314.3 | XP_006713377.1 | 5 | 7490 | ||
XM_011534041.2 | XP_011532343.1 | 5 | 7327 | |||
XM_017007088.1 | XP_016862577.1 | 6 | 7603 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
