siRNA Details
Gene Symbol: BTD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006713314.3 | XP_006713377.1 | 5 | 8705 | ||
XM_011534041.2 | XP_011532343.1 | 5 | 8542 | |||
XM_017007088.1 | XP_016862577.1 | 6 | 8818 |
Gene Symbol: C12orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025908.1 | - | 2 | 1212 | ||
BC020885.1 | - | 2 | 1232 | 4665283 |
Gene Symbol: GNB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091332.1 | - | 1 | 278 |
Gene Symbol: GTF2IRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123716.1 | - | 1 | 229 | ||
AK124334.1 | - | 1 | 556 |
Gene Symbol: HSCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318314.1 | NP_001305243.1 | 5 | 791 | ||
NM_001318315.1 | NP_001305244.1 | 5 | 796 | |||
NM_001318316.1 | NP_001305245.1 | 6 | 690 | |||
NM_172002.4 | NP_741999.3 | 6 | 936 | |||
NR_134560.1 | 6 | 881 | ||||
XM_005261359.3 | XP_005261416.1 | 6 | 937 | |||
XM_017028619.1 | XP_016884108.1 | 6 | 907 | |||
XM_017028620.1 | XP_016884109.1 | 6 | 740 | |||
GenBank mRNA | BC000004.1 | - | 6 | 945 | ||
BC065569.1 | - | 6 | 909 | 6171959 |
Gene Symbol: LOC100131894 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092825.1 | - | 1 | 1933 |
Gene Symbol: LOC101927877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135634.1 | 2 | 877 | |||
GenBank mRNA | BC040294.1 | - | 2 | 879 |
Gene Symbol: RHBDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304746.1 | NP_001291675.1 | 8 | 1540 | ||
NM_017821.4 | NP_060291.2 | 8 | 1570 | |||
GenBank mRNA | BC137108.1 | - | 7 | 1419 | 9021105 | |
BC137110.1 | - | 7 | 1419 | 9021107 | ||
BC144476.1 | - | 7 | 1406 | |||
BE876592.1 | - | 1 | 176 |
Gene Symbol: SEC14L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC139912.1 | - | 12 | 1590 | 40147686 |
Gene Symbol: SSU72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK129493.1 | - | 1 | 813 |
Gene Symbol: SUV39H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193424.1 | NP_001180353.1 | 6 | 2980 | ||
NM_001193425.1 | NP_001180354.1 | 6 | 2938 | |||
NM_001193426.1 | NP_001180355.1 | 6 | 2440 | |||
NM_001193427.1 | NP_001180356.1 | 6 | 2398 | |||
NM_024670.3 | NP_078946.1 | 5 | 2925 | |||
NR_034181.1 | 5 | 2834 | ||||
XM_011519662.2 | XP_011517964.1 | 6 | 2969 | |||
GenBank mRNA | AK027067.1 | - | 5 | 2731 | ||
BC007754.2 | - | 5 | 2925 | 4109807 |
Gene Symbol: TLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537631.1 | - | 4 | 2594 |
Gene Symbol: VRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125747.1 | - | 1 | 2035 |
Gene Symbol: WDR73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090406.1 | - | 1 | 893 |
Gene Symbol: WDR90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093802.1 | - | 14 | 2190 | ||
AK126622.1 | - | 20 | 3446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
