siRNA Details
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127183.2 | NP_001120655.1 | 10 | 4053 | ||
NM_001202515.1 | NP_001189444.1 | 6 | 3119 | |||
NM_001202516.1 | NP_001189445.1 | 8 | 3920 | |||
NM_001202517.1 | NP_001189446.1 | 9 | 3607 | |||
NM_001202518.1 | NP_001189447.1 | 8 | 4182 | |||
NM_001202519.1 | NP_001189448.1 | 9 | 4446 | |||
NM_003879.5 | NP_003870.4 | 10 | 4025 | |||
XM_017005189.1 | XP_016860678.1 | 11 | 4920 | |||
XM_017005190.1 | XP_016860679.1 | 10 | 3889 | |||
XM_017005191.1 | XP_016860680.1 | 10 | 3968 | |||
XM_017005192.1 | XP_016860681.1 | 10 | 4855 | |||
XM_017005193.1 | XP_016860682.1 | 10 | 6872 | |||
XM_017005194.1 | XP_016860683.1 | 10 | 4728 | |||
XM_017005195.1 | XP_016860684.1 | 11 | 4066 | |||
XM_017005196.1 | XP_016860685.1 | 8 | 3691 |
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336879.1 | - | 1 | 379 | ||
AF336883.1 | - | 1 | 364 | |||
AF336886.1 | - | 1 | 352 | |||
AF338190.1 | - | 1 | 397 | |||
AF338195.1 | - | 1 | 415 | |||
AF338196.1 | - | 1 | 427 | |||
AF338198.1 | - | 1 | 415 | |||
AF469044.1 | - | 1 | 364 |
Gene Symbol: NEK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126330.1 | - | 9 | 2720 |
Gene Symbol: SPICE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832327.2 | - | 12 | 3200 |
Gene Symbol: SQSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094484.1 | - | 1 | 89 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
