siRNA Details
Gene Symbol: ARRDC1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122035.1 | 3 | 667 | |||
NR_122036.1 | 3 | 356 | ||||
GenBank mRNA | AF267857.1 | - | 3 | 585 | ||
BC011781.2 | - | 3 | 569 | |||
BC066646.1 | - | 3 | 374 | 5749061 | ||
BC117267.1 | - | 2 | 127 | 40125818 | ||
CX164735.1 | - | 3 | 356 |
Gene Symbol: AMZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011515151.2 | XP_011513453.1 | 6 | 3051 | ||
XM_017011774.1 | XP_016867263.1 | 6 | 2996 |
Gene Symbol: AP2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209422.1 | - | 3 | 1368 |
Gene Symbol: DENND5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308339.1 | NP_001295268.1 | 2 | 243 | ||
XM_017018899.1 | XP_016874388.1 | 1 | 65 |
Gene Symbol: DISC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164537.1 | NP_001158009.1 | 4 | 1219 |
Gene Symbol: LOC105374266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135537.1 | 2 | 1773 |
Gene Symbol: LOC284454 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036515.1 | 1 | 1013 | |||
GenBank mRNA | AL832183.1 | - | 1 | 1010 | ||
BX640708.1 | - | 1 | 1013 |
Gene Symbol: SIGLEC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098612.1 | NP_001092082.1 | 7 | 1857 | ||
XM_017026113.1 | XP_016881602.1 | 6 | 1545 | |||
GenBank mRNA | AY854038.1 | - | 7 | 1781 | ||
CA439587.1 | - | 1 | 72 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
