siRNA Details
Gene Symbol: AGAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077665.2 | NP_001071133.2 | 4 | 783 | ||
XM_017016268.1 | XP_016871757.1 | 4 | 504 | |||
XM_017016269.1 | XP_016871758.1 | 4 | 504 | |||
XM_017016270.1 | XP_016871759.1 | 3 | 738 | |||
XM_017016271.1 | XP_016871760.1 | 6 | 506 | |||
XM_017016272.1 | XP_016871761.1 | 5 | 436 | |||
XM_017016273.1 | XP_016871762.1 | 4 | 795 | |||
XM_017016274.1 | XP_016871763.1 | 2 | 60 |
Gene Symbol: AGAP12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029396.2 | 4 | 427 |
Gene Symbol: AGAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276343.2 | NP_001263272.2 | 4 | 511 | ||
NM_133446.3 | NP_597703.2 | 3 | 442 | |||
XM_011539269.2 | XP_011537571.1 | 2 | 65 | |||
XM_017015664.1 | XP_016871153.1 | 6 | 4373 | |||
XM_017015665.1 | XP_016871154.1 | 7 | 4241 | |||
XM_017015666.1 | XP_016871155.1 | 5 | 1276 | |||
XM_017015667.1 | XP_016871156.1 | 6 | 546 | |||
XM_017015668.1 | XP_016871157.1 | 3 | 162 | |||
XM_017015669.1 | XP_016871158.1 | 3 | 115 | |||
GenBank mRNA | AF411132.1 | - | 3 | 442 | ||
BC171820.1 | - | 4 | 496 |
Gene Symbol: AGAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144000.1 | NP_001137472.1 | 4 | 426 |
Gene Symbol: AGAP7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126580.1 | 4 | 512 |
Gene Symbol: AGAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190810.1 | NP_001177739.1 | 4 | 497 | ||
GenBank mRNA | AK160375.1 | - | 5 | 3473 | ||
BC171845.1 | - | 4 | 497 |
Gene Symbol: LOC107984026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017029.1 | XP_016872518.1 | 4 | 823 | ||
XM_017017030.1 | XP_016872519.1 | 3 | 754 | |||
XM_017017031.1 | XP_016872520.1 | 3 | 754 | |||
XM_017017032.1 | XP_016872521.1 | 5 | 854 | |||
XM_017017033.1 | XP_016872522.1 | 6 | 617 | |||
XM_017017034.1 | XP_016872523.1 | 5 | 522 | |||
XM_017017037.1 | XP_016872526.1 | 4 | 791 | |||
XM_017017038.1 | XP_016872527.1 | 2 | 44 | |||
XM_017017040.1 | XP_016872529.1 | 3 | 123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
