siRNA Details
Gene Symbol: LOC107984026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017029.1 | XP_016872518.1 | 3 | 751 | ||
XM_017017030.1 | XP_016872519.1 | 2 | 682 | |||
XM_017017031.1 | XP_016872520.1 | 5 | 822 | |||
XM_017017032.1 | XP_016872521.1 | 4 | 782 | |||
XM_017017033.1 | XP_016872522.1 | 5 | 545 | |||
XM_017017034.1 | XP_016872523.1 | 4 | 450 | |||
XM_017017035.1 | XP_016872524.1 | 3 | 103 | |||
XM_017017037.1 | XP_016872526.1 | 3 | 719 | |||
XM_017017039.1 | XP_016872528.1 | 3 | 122 | |||
XM_017017040.1 | XP_016872529.1 | 2 | 51 |
Gene Symbol: AGAP12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029396.2 | 3 | 355 |
Gene Symbol: AGAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276343.2 | NP_001263272.2 | 3 | 439 | ||
NM_001291379.1 | NP_001278308.1 | 6 | 666 | |||
NM_133446.3 | NP_597703.2 | 2 | 370 | |||
XM_017015664.1 | XP_016871153.1 | 5 | 4301 | |||
XM_017015665.1 | XP_016871154.1 | 6 | 4169 | |||
XM_017015666.1 | XP_016871155.1 | 4 | 1204 | |||
XM_017015667.1 | XP_016871156.1 | 5 | 474 | |||
XM_017015668.1 | XP_016871157.1 | 2 | 90 | |||
XM_017015669.1 | XP_016871158.1 | 2 | 43 | |||
GenBank mRNA | AF411132.1 | - | 2 | 370 | ||
BC171820.1 | - | 3 | 424 |
Gene Symbol: AGAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144000.1 | NP_001137472.1 | 3 | 354 |
Gene Symbol: AGAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077665.2 | NP_001071133.2 | 3 | 711 | ||
XM_017016268.1 | XP_016871757.1 | 3 | 432 | |||
XM_017016270.1 | XP_016871759.1 | 2 | 666 | |||
XM_017016271.1 | XP_016871760.1 | 5 | 434 | |||
XM_017016272.1 | XP_016871761.1 | 4 | 364 | |||
XM_017016273.1 | XP_016871762.1 | 3 | 723 |
Gene Symbol: AGAP7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126580.1 | 3 | 440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
