siRNA Details
Gene Symbol: CYP2B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000767.4 | NP_000758.1 | 9 | 1811 | ||
XM_011526548.2 | XP_011524850.1 | 6 | 1424 | |||
XM_011526549.2 | XP_011524851.1 | 7 | 1366 | |||
XM_011526550.2 | XP_011524852.1 | 5 | 1219 | |||
GenBank mRNA | AK301620.1 | - | 5 | 1208 | ||
M29874.1 | - | 9 | 1810 | |||
X06399.1 | - | 3 | 1156 | |||
X06400.1 | - | 3 | 755 |
Gene Symbol: CAPZA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ655956.2 | - | 2 | 164 |
Gene Symbol: MRPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209905.1 | - | 5 | 3959 |
Gene Symbol: SLC25A45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077241.2 | NP_001070709.2 | 6 | 1459 | ||
NM_001278250.2 | NP_001265179.2 | 5 | 1670 | |||
NM_001278251.2 | NP_001265180.2 | 10 | 2316 | |||
NM_001300820.1 | NP_001287749.1 | 5 | 1556 | |||
NM_182556.3 | NP_872362.3 | 7 | 1514 | |||
XM_006718509.3 | XP_006718572.1 | 6 | 2124 | |||
XM_006718510.3 | XP_006718573.1 | 8 | 1652 | |||
XM_011544943.2 | XP_011543245.1 | 7 | 2223 | |||
XM_011544944.2 | XP_011543246.1 | 6 | 1553 | |||
XM_011544949.2 | XP_011543251.1 | 8 | 1571 | |||
XM_017017563.1 | XP_016873052.1 | 7 | 2989 | |||
XM_017017564.1 | XP_016873053.1 | 7 | 2988 | |||
XM_017017565.1 | XP_016873054.1 | 7 | 2253 | |||
XM_017017566.1 | XP_016873055.1 | 7 | 1608 | |||
XM_017017567.1 | XP_016873056.1 | 8 | 1614 | |||
XM_017017568.1 | XP_016873057.1 | 7 | 1437 | |||
XM_017017569.1 | XP_016873058.1 | 9 | 2316 | |||
XM_017017570.1 | XP_016873059.1 | 5 | 1939 | |||
XM_017017571.1 | XP_016873060.1 | 5 | 1309 | |||
GenBank mRNA | AK090434.1 | - | 2 | 3570 | ||
AK091190.1 | - | 10 | 2316 | |||
BC036869.1 | - | 6 | 1426 | 5228549 | ||
BX648573.1 | - | 3 | 6343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
