siRNA Details
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130464.2 | NP_569731.2 | 4 | 347 | ||
GenBank mRNA | AB209632.1 | - | 3 | 2914 | ||
AK295170.1 | - | 4 | 346 | |||
AK304162.1 | - | 2 | 129 | |||
BC036263.1 | - | 4 | 347 | 3903371 | ||
BC094882.1 | - | 3 | 274 | 3093162 | ||
DC342292.1 | - | 3 | 451 | |||
KF175175.1 | - | 3 | 214 | |||
KF175176.1 | - | 3 | 271 | |||
KF175177.1 | - | 3 | 271 | |||
KF175178.1 | - | 3 | 172 | |||
KF175179.1 | - | 3 | 214 | |||
KF175180.1 | - | 3 | 214 | |||
KF175181.1 | - | 3 | 358 | |||
KF175182.1 | - | 3 | 214 | |||
KF175183.1 | - | 2 | 106 | |||
KF175184.1 | - | 2 | 97 | |||
KF175185.1 | - | 3 | 214 | |||
KF175186.1 | - | 2 | 88 | |||
KF175187.1 | - | 3 | 214 | |||
KF175188.1 | - | 3 | 175 | |||
KF175189.1 | - | 3 | 175 | |||
KF175190.1 | - | 3 | 214 | |||
KF175191.1 | - | 3 | 175 | |||
KF175192.1 | - | 3 | 175 | |||
KF175193.1 | - | 3 | 178 | |||
KF175194.1 | - | 3 | 178 | |||
KF175196.1 | - | 3 | 175 | |||
KF175197.1 | - | 3 | 175 | |||
KF175198.1 | - | 3 | 271 | |||
KF175199.1 | - | 3 | 271 | |||
KF175200.1 | - | 3 | 271 | |||
KF175201.1 | - | 3 | 214 | |||
KF175202.1 | - | 3 | 175 | |||
KF175203.1 | - | 3 | 271 | |||
KF175205.1 | - | 3 | 271 | |||
KF175206.1 | - | 3 | 271 | |||
KF175215.1 | - | 5 | 533 | |||
KF175216.1 | - | 4 | 548 | |||
KF175218.1 | - | 3 | 176 | |||
KF175223.1 | - | 3 | 271 | |||
KF175224.1 | - | 3 | 214 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321892.1 | NP_001308821.1 | 5 | 464 | ||
XM_017023514.1 | XP_016879003.1 | 5 | 753 | |||
XM_017023515.1 | XP_016879004.1 | 6 | 772 | |||
XM_017023516.1 | XP_016879005.1 | 5 | 799 | |||
XM_017023517.1 | XP_016879006.1 | 5 | 530 | |||
XM_017023518.1 | XP_016879007.1 | 5 | 618 | |||
XM_017023519.1 | XP_016879008.1 | 5 | 675 | |||
GenBank mRNA | BQ433543.1 | - | 387 | |||
DB231720.1 | - | 4 | 333 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310137.1 | NP_001297066.1 | 3 | 271 | ||
GenBank mRNA | BF592449.1 | - | 3 | 257 | ||
KF175195.1 | - | 3 | 175 | |||
KF175204.1 | - | 3 | 214 |
Gene Symbol: NPIPB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306094.1 | NP_001293023.1 | 3 | 293 | ||
XM_011523496.2 | XP_011521798.1 | 4 | 349 | |||
XM_011523498.2 | XP_011521800.1 | 4 | 383 | |||
XM_017023248.1 | XP_016878737.1 | 4 | 786 | |||
XM_017023249.1 | XP_016878738.1 | 3 | 603 |
Gene Symbol: NPIPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310148.1 | NP_001297077.1 | 4 | 496 | ||
XM_017023244.1 | XP_016878733.1 | 3 | 2507 | |||
XM_017023245.1 | XP_016878734.1 | 4 | 2344 | |||
XM_017023246.1 | XP_016878735.1 | 4 | 2265 | |||
XM_017023247.1 | XP_016878736.1 | 3 | 498 |
Gene Symbol: NPIPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135865.1 | NP_001129337.1 | 3 | 550 | ||
GenBank mRNA | AK296338.1 | - | 3 | 391 |
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282524.1 | NP_001269453.1 | 3 | 363 | ||
XM_005255741.3 | XP_005255798.1 | 4 | 1175 | |||
XM_005255742.4 | XP_005255799.1 | 4 | 921 | |||
XM_011545929.2 | XP_011544231.1 | 5 | 995 | |||
XM_011545931.2 | XP_011544233.1 | 4 | 628 | |||
XM_017023628.1 | XP_016879117.1 | 4 | 378 | |||
XM_017023629.1 | XP_016879118.1 | 4 | 921 | |||
XM_017023630.1 | XP_016879119.1 | 5 | 648 | |||
XM_017023631.1 | XP_016879120.1 | 3 | 992 | |||
XM_017023632.1 | XP_016879121.1 | 4 | 378 | |||
GenBank mRNA | BC144439.1 | - | 3 | 324 | ||
BC157836.1 | - | 3 | 1013 | 9057055 | ||
KF175222.1 | - | 3 | 325 |
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310136.1 | NP_001297065.1 | 3 | 325 | ||
XM_017023622.1 | XP_016879111.1 | 4 | 449 | |||
XM_017023623.1 | XP_016879112.1 | 4 | 449 | |||
XM_017023624.1 | XP_016879113.1 | 4 | 449 | |||
XM_017023625.1 | XP_016879114.1 | 4 | 429 | |||
XM_017023626.1 | XP_016879115.1 | 5 | 1068 | |||
XM_017023627.1 | XP_016879116.1 | 4 | 449 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287250.1 | NP_001274179.1 | 3 | 363 | ||
NM_001287251.1 | NP_001274180.1 | 4 | 449 | |||
XM_003118719.7 | XP_003118767.2 | 4 | 924 | |||
XM_011545690.2 | XP_011543992.1 | 5 | 1013 | |||
XM_011545693.2 | XP_011543995.1 | 5 | 1012 | |||
XM_017022814.1 | XP_016878303.1 | 5 | 1013 | |||
XM_017022815.1 | XP_016878304.1 | 5 | 533 | |||
XM_017022816.1 | XP_016878305.1 | 5 | 1013 | |||
XM_017022817.1 | XP_016878306.1 | 4 | 925 | |||
XM_017022818.1 | XP_016878307.1 | 4 | 2250 | |||
XM_017022819.1 | XP_016878308.1 | 5 | 940 | |||
XM_017022820.1 | XP_016878309.1 | 5 | 657 | |||
XM_017022821.1 | XP_016878310.1 | 4 | 1523 | |||
XM_017022822.1 | XP_016878311.1 | 5 | 2526 | |||
XM_017022823.1 | XP_016878312.1 | 3 | 2162 | |||
XM_017022824.1 | XP_016878313.1 | 5 | 939 | |||
XM_017022825.1 | XP_016878314.1 | 5 | 657 | |||
XM_017022826.1 | XP_016878315.1 | 4 | 474 | |||
XM_017022828.1 | XP_016878317.1 | 5 | 1012 | |||
GenBank mRNA | BC171825.1 | - | 4 | 1099 |
Gene Symbol: PDXDC2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003610.1 | 22 | 2296 | |||
GenBank mRNA | AL833251.1 | - | 22 | 2296 | ||
BC063633.1 | - | 20 | 1915 | |||
BC144350.1 | - | 3 | 271 | 9052870 | ||
KF175207.1 | - | 3 | 271 | |||
KF175220.1 | - | 3 | 176 | |||
KF175221.1 | - | 3 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
