siRNA Details
Gene Symbol: H3F3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002107.4 | NP_002098.1 | 2 | 246 | ||
GenBank mRNA | AK293541.1 | - | 2 | 228 | ||
AK311905.1 | - | 2 | 246 | |||
BC013857.1 | - | 1 | 114 | |||
BC029405.1 | - | 2 | 228 | 4077662 | ||
BC038989.1 | - | 1 | 148 | 6014954 | ||
BC081561.1 | - | 1 | 219 | 5162817 | ||
BC095447.1 | - | 2 | 228 | 30718542 | ||
BM760856.1 | - | 2 | 233 | |||
M11353.1 | - | 1 | 492 | |||
M11354.1 | - | 2 | 226 |
Gene Symbol: H3F3AP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002315.1 | 1 | 226 | |||
GenBank mRNA | AK291102.1 | - | 2 | 228 | ||
BC066901.1 | - | 1 | 222 | |||
BC067757.1 | - | 1 | 228 | |||
BC081560.1 | - | 1 | 219 | 3884607 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
