siRNA Details
Gene Symbol: SPANXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013453.3 | NP_038481.2 | 2 | 247 | ||
GenBank mRNA | AF098306.1 | - | 2 | 223 | ||
AY920732.1 | - | 2 | 187 | |||
AY920733.1 | - | 2 | 187 | |||
AY920735.1 | - | 2 | 187 | |||
AY920736.1 | - | 2 | 187 | |||
AY920738.1 | - | 2 | 187 | |||
AY920739.1 | - | 2 | 187 | |||
AY920741.1 | - | 2 | 187 | |||
AY920742.1 | - | 2 | 187 | |||
AY920743.1 | - | 2 | 187 | |||
AY920744.1 | - | 2 | 187 | |||
AY920745.1 | - | 2 | 187 | |||
AY920746.1 | - | 2 | 187 | |||
BC069393.1 | - | 2 | 223 | 7262682 | ||
BC069816.1 | - | 2 | 223 | 7262670 | ||
CB115288.1 | - | 2 | 247 |
Gene Symbol: SPANXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145662.3 | NP_663695.1 | 2 | 247 | ||
GenBank mRNA | CB110020.1 | - | 2 | 247 | ||
CB113891.1 | - | 2 | 247 |
Gene Symbol: SPANXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032417.3 | NP_115793.1 | 2 | 521 | ||
GenBank mRNA | AF312765.1 | - | 2 | 197 | ||
AY920734.1 | - | 2 | 187 | |||
AY920740.1 | - | 2 | 187 | |||
BC103961.1 | - | 2 | 220 | 40013988 | ||
BC121070.2 | - | 2 | 230 | 40119569 | ||
BC134418.1 | - | 2 | 230 | 40119568 | ||
BM554791.1 | - | 2 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
