siRNA Details
Gene Symbol: IGHV3-69-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF460695.1 | - | 67 | |||
AF460697.1 | - | 67 | ||||
AF460787.1 | - | 67 | ||||
GU552754.1 | - | 124 | ||||
JX420391.1 | - | 1 | 109 | |||
JX420392.1 | - | 1 | 109 |
Gene Symbol: IGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX640854.1 | - | 2 | 351 |
Gene Symbol: IGHG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC062335.1 | - | 2 | 334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
