siRNA Details
Gene Symbol: RBMY1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303410.1 | NP_001290339.1 | 10 | 1188 | ||
NM_152585.2 | NP_689798.1 | 10 | 1188 | |||
XM_017030032.1 | XP_016885521.1 | 7 | 777 | |||
GenBank mRNA | BC030018.2 | - | 10 | 1188 | 5269012 | |
X76060.1 | - | 10 | 1170 |
Gene Symbol: RBMY1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320945.1 | NP_001307874.1 | 9 | 1050 | ||
NM_005058.3 | NP_005049.1 | 10 | 1228 | |||
XM_011531437.2 | XP_011529739.1 | 10 | 1167 | |||
GenBank mRNA | BC047768.1 | - | 9 | 1025 | 5744153 | |
BC146853.1 | - | 10 | 1141 | 9056781 | ||
X76059.1 | - | 10 | 1193 |
Gene Symbol: RBMY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006121.3 | NP_001006121.1 | 10 | 1228 | ||
XM_011531434.1 | XP_011529736.1 | 9 | 1027 | |||
XM_017030056.1 | XP_016885545.1 | 10 | 1188 |
Gene Symbol: RBMY1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006120.3 | NP_001006120.2 | 10 | 1228 | ||
XM_011531486.2 | XP_011529788.1 | 10 | 1101 | |||
XM_011531487.2 | XP_011529789.1 | 10 | 1188 | |||
XM_011531488.2 | XP_011529790.1 | 10 | 1188 | |||
XM_011531490.1 | XP_011529792.1 | 9 | 1027 |
Gene Symbol: RBMY1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006118.2 | NP_001006118.2 | 10 | 1188 | ||
XM_011531494.2 | XP_011529796.1 | 10 | 1188 | |||
XM_011531496.1 | XP_011529798.1 | 9 | 1027 |
Gene Symbol: RBMY1J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006117.3 | NP_001006117.2 | 10 | 1253 | ||
XM_011531503.1 | XP_011529805.1 | 7 | 777 | |||
GenBank mRNA | AK313597.1 | - | 10 | 1257 | ||
BC144577.1 | - | 10 | 1141 | 9053106 | ||
BC152392.1 | - | 9 | 1098 | 8860211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
