siRNA Details
Gene Symbol: ME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168335.1 | NP_001161807.1 | 14 | 3718 | ||
NM_002396.4 | NP_002387.1 | 16 | 3888 |
Gene Symbol: MIR646HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128288.1 | - | 1 | 2754 |
Gene Symbol: PDE12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322176.1 | NP_001309105.1 | 3 | 2150 |
Gene Symbol: RHNO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252499.2 | NP_001239428.1 | 3 | 1174 | ||
NM_001252500.2 | NP_001239429.1 | 3 | 1132 | |||
NM_001257097.1 | NP_001244026.1 | 3 | 1295 | |||
NM_001257098.1 | NP_001244027.1 | 3 | 1194 | |||
NR_027365.2 | 2 | 922 | ||||
NR_046432.1 | 4 | 1308 | ||||
NR_046433.1 | 2 | 1096 | ||||
GenBank mRNA | AB073599.1 | - | 3 | 1150 | ||
AK021945.1 | - | 2 | 898 | |||
AK304613.1 | - | 3 | 1108 | |||
BC005106.1 | - | 3 | 1140 |
Gene Symbol: ZNF619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145082.2 | NP_001138554.1 | 6 | 2795 | ||
NM_001145093.2 | NP_001138565.1 | 5 | 2675 | |||
NR_026746.2 | 6 | 2825 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
