siRNA Details
Gene Symbol: MAGEA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282501.1 | NP_001269430.1 | 5 | 754 | ||
NM_001282502.1 | NP_001269431.1 | 4 | 656 | |||
NM_001282504.1 | NP_001269433.1 | 3 | 536 | |||
NM_001282505.1 | NP_001269434.1 | 2 | 470 | |||
NM_005361.3 | NP_005352.1 | 4 | 831 | |||
NM_175742.2 | NP_786884.1 | 6 | 817 | |||
NM_175743.2 | NP_786885.1 | 6 | 833 | |||
XM_017029519.1 | XP_016885008.1 | 5 | 793 | |||
GenBank mRNA | AK290443.1 | - | 5 | 754 | ||
BC013098.2 | - | 2 | 440 | 4343986 | ||
BC063681.1 | - | 4 | 620 | 4403536 | ||
BC108720.1 | - | 4 | 650 | 6065905 | ||
BC109188.1 | - | 2 | 393 | |||
BC112158.1 | - | 1 | 338 | 8327626 | ||
BC112160.1 | - | 1 | 338 | 8327628 | ||
DR158683.1 | - | 3 | 521 |
Gene Symbol: MAGEA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321400.1 | NP_001308329.1 | 6 | 833 | ||
NM_001321401.1 | NP_001308330.1 | 6 | 817 | |||
NM_001321402.1 | NP_001308331.1 | 5 | 754 | |||
NM_001321403.1 | NP_001308332.1 | 4 | 656 | |||
NM_001321404.1 | NP_001308333.1 | 2 | 470 | |||
NM_153488.4 | NP_705692.1 | 3 | 536 | |||
XM_017029406.1 | XP_016884895.1 | 5 | 792 |
Gene Symbol: MAGEA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005362.3 | NP_005353.1 | 3 | 517 | ||
XM_005274676.3 | XP_005274733.1 | 3 | 591 | |||
XM_006724818.2 | XP_006724881.1 | 3 | 673 | |||
XM_011531160.2 | XP_011529462.1 | 3 | 497 | |||
XM_011531161.2 | XP_011529463.1 | 3 | 922 | |||
GenBank mRNA | AK223519.1 | - | 1 | 344 | ||
AK292384.1 | - | 3 | 517 | |||
BC000340.2 | - | 3 | 482 | 2822978 | ||
BC005963.1 | - | 3 | 517 | 4064879 | ||
BC011744.2 | - | 2 | 443 | 3345801 | ||
BC016803.1 | - | 3 | 479 | 4073332 | ||
BC017389.1 | - | 3 | 474 | 4047542 | ||
CR541767.1 | - | 1 | 308 | |||
CR541774.1 | - | 1 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
