siRNA Details
Gene Symbol: MAGEA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005363.3 | NP_005354.1 | 3 | 569 | ||
NM_175868.2 | NP_787064.1 | 3 | 633 | |||
GenBank mRNA | AK289437.1 | - | 3 | 553 | ||
BC041599.1 | - | 3 | 542 | 5458512 | ||
BC067731.1 | - | 3 | 550 | 4828429 | ||
BM015284.1 | - | 3 | 569 | |||
BU182516.1 | - | 3 | 583 | |||
BX640600.1 | - | 3 | 930 | |||
D32076.1 | - | 1 | 427 | |||
EU161101.1 | - | 1 | 362 | |||
U10339.1 | - | 1 | 362 |
Gene Symbol: MAGEA10-MAGEA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204811.2 | NP_001191740.1 | 4 | 743 |
Gene Symbol: MAGEA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166386.3 | NP_001159858.1 | 3 | 716 | ||
NM_001166387.3 | NP_001159859.1 | 3 | 624 | |||
NM_005367.6 | NP_005358.2 | 2 | 518 | |||
GenBank mRNA | AK314168.1 | - | 3 | 716 | ||
BC003408.2 | - | 2 | 506 | 3450217 | ||
BC098146.1 | - | 3 | 296 | |||
BQ436888.1 | - | 2 | 518 | |||
BT007108.1 | - | 1 | 362 | |||
CR541775.1 | - | 1 | 362 |
Gene Symbol: MAGEA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282501.1 | NP_001269430.1 | 5 | 808 | ||
NM_001282502.1 | NP_001269431.1 | 4 | 710 | |||
NM_001282504.1 | NP_001269433.1 | 3 | 590 | |||
NM_001282505.1 | NP_001269434.1 | 2 | 524 | |||
NM_005361.3 | NP_005352.1 | 4 | 885 | |||
NM_175742.2 | NP_786884.1 | 6 | 871 | |||
NM_175743.2 | NP_786885.1 | 6 | 887 | |||
XM_017029519.1 | XP_016885008.1 | 5 | 847 | |||
GenBank mRNA | AK290443.1 | - | 5 | 808 | ||
BC013098.2 | - | 2 | 494 | 4343986 | ||
BC063681.1 | - | 4 | 674 | 4403536 | ||
BC108720.1 | - | 4 | 704 | 6065905 | ||
BC109188.1 | - | 2 | 447 | |||
BC112158.1 | - | 1 | 392 | 8327626 | ||
BC112160.1 | - | 1 | 392 | 8327628 | ||
DR158683.1 | - | 3 | 575 |
Gene Symbol: MAGEA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321400.1 | NP_001308329.1 | 6 | 887 | ||
NM_001321401.1 | NP_001308330.1 | 6 | 871 | |||
NM_001321402.1 | NP_001308331.1 | 5 | 808 | |||
NM_001321403.1 | NP_001308332.1 | 4 | 710 | |||
NM_001321404.1 | NP_001308333.1 | 2 | 524 | |||
NM_153488.4 | NP_705692.1 | 3 | 590 | |||
XM_017029406.1 | XP_016884895.1 | 5 | 846 |
Gene Symbol: MAGEA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005362.3 | NP_005353.1 | 3 | 571 | ||
XM_005274676.3 | XP_005274733.1 | 3 | 645 | |||
XM_006724818.2 | XP_006724881.1 | 3 | 727 | |||
XM_011531160.2 | XP_011529462.1 | 3 | 551 | |||
XM_011531161.2 | XP_011529463.1 | 3 | 976 | |||
GenBank mRNA | AK223519.1 | - | 1 | 398 | ||
AK292384.1 | - | 3 | 571 | |||
BC000340.2 | - | 3 | 536 | 2822978 | ||
BC005963.1 | - | 3 | 571 | 4064879 | ||
BC011744.2 | - | 2 | 497 | 3345801 | ||
BC016803.1 | - | 3 | 533 | 4073332 | ||
BC017389.1 | - | 3 | 528 | 4047542 | ||
CR541767.1 | - | 1 | 362 | |||
CR541768.1 | - | 1 | 362 | |||
CR541774.1 | - | 1 | 362 | |||
CR542020.1 | - | 1 | 362 |
Gene Symbol: MAGEA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021049.4 | NP_066387.1 | 3 | 534 | ||
GenBank mRNA | AY310324.1 | - | 1 | 362 | ||
BC109187.1 | - | 3 | 534 | 40000127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
