siRNA Details
Gene Symbol: MAGEA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005363.3 | NP_005354.1 | 3 | 1055 | ||
NM_175868.2 | NP_787064.1 | 3 | 1119 | |||
GenBank mRNA | AK289437.1 | - | 3 | 1039 | ||
BC041599.1 | - | 3 | 1028 | 5458512 | ||
BC067731.1 | - | 3 | 1036 | 4828429 | ||
BX640600.1 | - | 3 | 1416 | |||
D32076.1 | - | 1 | 913 | |||
EU161101.1 | - | 1 | 848 | |||
U10339.1 | - | 1 | 848 |
Gene Symbol: MAGEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004988.4 | NP_004979.3 | 3 | 1047 | ||
GenBank mRNA | AK314979.1 | - | 3 | 1047 | ||
AY148486.1 | - | 3 | 1005 | |||
BC017555.1 | - | 3 | 1014 | 3922845 | ||
BT009789.1 | - | 1 | 827 |
Gene Symbol: MAGEA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166386.3 | NP_001159858.1 | 3 | 1202 | ||
NM_001166387.3 | NP_001159859.1 | 3 | 1110 | |||
NM_005367.6 | NP_005358.2 | 2 | 1004 | |||
GenBank mRNA | AK314168.1 | - | 3 | 1202 | ||
BC003408.2 | - | 2 | 992 | 3450217 | ||
BT007108.1 | - | 1 | 848 | |||
CR541775.1 | - | 1 | 848 |
Gene Symbol: MAGEA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282501.1 | NP_001269430.1 | 5 | 1294 | ||
NM_001282502.1 | NP_001269431.1 | 4 | 1196 | |||
NM_001282504.1 | NP_001269433.1 | 3 | 1076 | |||
NM_001282505.1 | NP_001269434.1 | 2 | 1010 | |||
NM_005361.3 | NP_005352.1 | 4 | 1371 | |||
NM_175742.2 | NP_786884.1 | 6 | 1357 | |||
NM_175743.2 | NP_786885.1 | 6 | 1373 | |||
XM_017029519.1 | XP_016885008.1 | 5 | 1333 | |||
GenBank mRNA | AK290443.1 | - | 5 | 1294 | ||
BC013098.2 | - | 2 | 980 | 4343986 | ||
BC063681.1 | - | 4 | 1160 | 4403536 | ||
BC108720.1 | - | 4 | 1190 | 6065905 | ||
BC112158.1 | - | 1 | 878 | 8327626 | ||
BC112160.1 | - | 1 | 878 | 8327628 |
Gene Symbol: MAGEA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321400.1 | NP_001308329.1 | 6 | 1373 | ||
NM_001321401.1 | NP_001308330.1 | 6 | 1357 | |||
NM_001321402.1 | NP_001308331.1 | 5 | 1294 | |||
NM_001321403.1 | NP_001308332.1 | 4 | 1196 | |||
NM_001321404.1 | NP_001308333.1 | 2 | 1010 | |||
NM_153488.4 | NP_705692.1 | 3 | 1076 | |||
XM_017029406.1 | XP_016884895.1 | 5 | 1332 |
Gene Symbol: MAGEA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005362.3 | NP_005353.1 | 3 | 1057 | ||
XM_005274676.3 | XP_005274733.1 | 3 | 1131 | |||
XM_006724818.2 | XP_006724881.1 | 3 | 1213 | |||
XM_011531160.2 | XP_011529462.1 | 3 | 1037 | |||
XM_011531161.2 | XP_011529463.1 | 3 | 1462 | |||
GenBank mRNA | AK223519.1 | - | 1 | 884 | ||
AK292384.1 | - | 3 | 1057 | |||
BC000340.2 | - | 3 | 1022 | 2822978 | ||
BC005963.1 | - | 3 | 1057 | 4064879 | ||
BC011744.2 | - | 2 | 983 | 3345801 | ||
BC016803.1 | - | 3 | 1019 | 4073332 | ||
BC017389.1 | - | 3 | 1014 | 4047542 | ||
CR541767.1 | - | 1 | 848 | |||
CR541768.1 | - | 1 | 848 | |||
CR541774.1 | - | 1 | 848 |
Gene Symbol: MAGEA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011548.1 | NP_001011548.1 | 3 | 1069 | ||
NM_001011549.1 | NP_001011549.1 | 3 | 1066 | |||
NM_001011550.1 | NP_001011550.1 | 3 | 1057 | |||
NM_002362.4 | NP_002353.3 | 3 | 1072 | |||
XM_005274677.2 | XP_005274734.1 | 3 | 1112 | |||
XM_005274678.3 | XP_005274735.1 | 3 | 1070 | |||
XM_005274679.3 | XP_005274736.1 | 3 | 1079 | |||
GenBank mRNA | AK300179.1 | - | 2 | 1129 | ||
AK313862.1 | - | 3 | 1108 | |||
AY954624.1 | - | 2 | 942 | |||
BC017723.1 | - | 3 | 1069 | 4394783 | ||
D32075.1 | - | 1 | 916 | |||
D32077.1 | - | 1 | 916 | |||
U10340.1 | - | 1 | 851 |
Gene Symbol: MAGEA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166400.1 | NP_001159872.1 | 4 | 1406 | ||
NM_001166401.1 | NP_001159873.1 | 3 | 1378 | |||
NM_005364.4 | NP_005355.2 | 3 | 1127 | |||
XM_017029520.1 | XP_016885009.1 | 4 | 1194 | |||
GenBank mRNA | AK313943.1 | - | 3 | 1127 | ||
BC002455.2 | - | 3 | 1073 | 3345962 | ||
BC012744.2 | - | 3 | 1066 | 3617812 | ||
BT007340.1 | - | 1 | 857 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
