siRNA Details
Gene Symbol: HMGN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005517.3 | NP_005508.1 | 4 | 316 | ||
GenBank mRNA | AK026562.1 | - | 4 | 158 | ||
BC000378.1 | - | 4 | 219 | 2820349 | ||
BC003050.1 | - | 4 | 209 | 3543809 | ||
BC003689.1 | - | 4 | 180 | |||
BC014644.1 | - | 4 | 199 | 3355317 | ||
BC032140.2 | - | 4 | 208 | 5021546 | ||
BC070297.1 | - | 2 | 167 | 4282630 | ||
BC071707.1 | - | 4 | 209 | 3849426 | ||
BC072010.1 | - | 4 | 203 | 6273507 | ||
BC072011.1 | - | 4 | 199 | 6647282 | ||
BC075837.1 | - | 4 | 304 | 6272425 | ||
BC081567.1 | - | 4 | 202 | 4131303 | ||
BC110390.1 | - | 4 | 212 | 5190365 | ||
CR542122.1 | - | 1 | 114 | |||
M12623.1 | - | 4 | 202 |
Gene Symbol: TRMT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020810.3 | NP_065861.3 | 5 | 4392 | ||
XM_005267916.3 | XP_005267973.2 | 5 | 4422 | |||
GenBank mRNA | AB037814.1 | - | 4 | 4264 | ||
BC063551.1 | - | 1 | 447 | |||
BC136606.1 | - | 5 | 4307 | 9020596 | ||
BC136607.1 | - | 5 | 4307 | 9020597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
