siRNA Details
Gene Symbol: ERVW-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130925.1 | NP_001124397.1 | 2 | 1724 | ||
NM_014590.3 | NP_055405.3 | 1 | 2006 | |||
GenBank mRNA | AF072505.1 | - | 2 | 1681 | ||
AF072506.2 | - | 2 | 1724 | |||
AF072508.1 | - | 1 | 77 | |||
AF208161.1 | - | 1 | 1892 | |||
AF506835.1 | - | 1 | 987 | |||
AF513360.1 | - | 1 | 1021 | |||
BC137381.1 | - | 1 | 1623 | 9021379 | ||
GQ919057.1 | - | 1 | 963 |
Gene Symbol: LOC105377310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011532442.2 | XP_011530744.1 | 1 | 181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
