siRNA Details
Gene Symbol: ERVW-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130925.1 | NP_001124397.1 | 2 | 1901 | ||
NM_014590.3 | NP_055405.3 | 1 | 2183 | |||
GenBank mRNA | AF072505.1 | - | 2 | 1858 | ||
AF072506.2 | - | 2 | 1901 | |||
AF072508.1 | - | 1 | 254 | |||
AF208161.1 | - | 1 | 2069 | |||
AF506835.1 | - | 1 | 1164 | |||
AF513360.1 | - | 1 | 1198 | |||
BC137381.1 | - | 1 | 1800 | 9021379 | ||
GQ919057.1 | - | 1 | 1140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
