siRNA Details
Gene Symbol: SMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034021.1 | 6 | 604 | |||
GenBank mRNA | AK289851.1 | - | 5 | 530 | ||
CN312642.1 | - | 3 | 394 | |||
X83301.1 | - | 6 | 604 |
Gene Symbol: GUSBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033968.1 | 3 | 342 | |||
GenBank mRNA | AW500063.1 | - | 3 | 275 | ||
BC104473.1 | - | 3 | 342 | |||
CF141910.1 | - | 3 | 348 |
Gene Symbol: SMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094021.1 | - | 6 | 933 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
