siRNA Details
Gene Symbol: ABCC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020297.3 | NP_064693.2 | 38 | 7087 | ||
XM_005253284.3 | XP_005253341.1 | 40 | 7873 | |||
XM_005253286.3 | XP_005253343.1 | 41 | 7933 | |||
XM_005253287.4 | XP_005253344.1 | 41 | 8049 | |||
XM_005253288.3 | XP_005253345.1 | 39 | 7757 | |||
XM_005253289.3 | XP_005253346.1 | 39 | 7834 | |||
XM_005253290.3 | XP_005253347.1 | 38 | 7732 | |||
XM_006719025.3 | XP_006719088.1 | 40 | 8010 | |||
XM_011520545.2 | XP_011518847.1 | 40 | 7929 | |||
GenBank mRNA | BX640893.1 | - | 1 | 2022 |
Gene Symbol: ATP9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC053561.1 | - | 5 | 1456 |
Gene Symbol: CFAP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC041426.1 | - | 7 | 1780 |
Gene Symbol: DDX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209392.1 | - | 4 | 2710 |
Gene Symbol: DTNB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003536.1 | XP_016859025.1 | 16 | 2417 | ||
GenBank mRNA | Y15718.1 | - | 17 | 2392 |
Gene Symbol: FLJ22447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026100.1 | - | 5 | 689 |
Gene Symbol: FNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC007861.1 | - | 3 | 676 |
Gene Symbol: FSIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092099.1 | - | 8 | 2543 |
Gene Symbol: LINC00504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126435.1 | 7 | 2979 | |||
GenBank mRNA | BC036758.2 | - | 7 | 2991 | 5563071 |
Gene Symbol: LINC00976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BF691581.1 | - | 501 |
Gene Symbol: LOC100129434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125368.1 | 3 | 2761 | |||
GenBank mRNA | AK096196.1 | - | 3 | 2970 |
Gene Symbol: LOC101926898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125837.1 | 3 | 1237 | |||
GenBank mRNA | BC042616.1 | - | 3 | 1239 |
Gene Symbol: LOC105372751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135520.1 | 6 | 1655 | |||
GenBank mRNA | AK124194.1 | - | 6 | 1655 |
Gene Symbol: LOC283728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095617.1 | - | 2 | 3379 |
Gene Symbol: SMC5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936772.1 | - | 3 | 6643 |
Gene Symbol: TCF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647333.1 | - | 1 | 6761 |
Gene Symbol: UGDH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047679.1 | 5 | 2862 | |||
GenBank mRNA | AK056558.1 | - | 5 | 2682 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
