siRNA Details
Gene Symbol: RPL23AP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_000029.3 | 3 | 603 | |||
NR_024528.1 | 4 | 854 | ||||
NR_024529.1 | 3 | 760 | ||||
GenBank mRNA | BC000596.1 | - | 3 | 554 | ||
BC105600.1 | - | 3 | 555 | |||
BX643196.1 | - | 3 | 576 |
Gene Symbol: CENPBD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC065480.1 | - | 4 | 854 |
Gene Symbol: RPL23AP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026981.1 | 4 | 918 | |||
NR_026982.1 | 3 | 846 | ||||
GenBank mRNA | AK293095.1 | - | 3 | 569 | ||
BC065556.1 | - | 3 | 713 | |||
BQ668440.1 | - | 4 | 918 |
Gene Symbol: RPL23AP87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029406.1 | 4 | 1756 | |||
GenBank mRNA | AK125669.1 | - | 4 | 1754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
