siRNA Details
Gene Symbol: RPL23AP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_000029.3 | 3 | 681 | |||
NR_024528.1 | 4 | 932 | ||||
NR_024529.1 | 3 | 838 | ||||
GenBank mRNA | BC000596.1 | - | 3 | 632 | ||
BC105600.1 | - | 3 | 633 | |||
BX643196.1 | - | 3 | 654 |
Gene Symbol: CENPBD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC065480.1 | - | 4 | 929 |
Gene Symbol: RPL23AP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026981.1 | 4 | 996 | |||
NR_026982.1 | 3 | 924 | ||||
GenBank mRNA | AK293095.1 | - | 3 | 647 | ||
BC065556.1 | - | 3 | 791 |
Gene Symbol: RPL23AP87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029406.1 | 4 | 1834 | |||
GenBank mRNA | AK125669.1 | - | 4 | 1832 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
