siRNA Details
Gene Symbol: RPL23AP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003572.2 | 4 | 751 | |||
GenBank mRNA | AK127000.1 | - | 3 | 480 | ||
BC052779.1 | - | 5 | 811 |
Gene Symbol: CENPBD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC065480.1 | - | 4 | 806 |
Gene Symbol: RPL23AP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_000029.3 | 3 | 555 | |||
NR_024528.1 | 4 | 806 | ||||
NR_024529.1 | 3 | 712 | ||||
GenBank mRNA | BC000596.1 | - | 3 | 506 | ||
BC105600.1 | - | 3 | 507 | |||
BG335018.1 | - | 4 | 746 | |||
BX643196.1 | - | 3 | 528 |
Gene Symbol: RPL23AP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026981.1 | 4 | 870 | |||
NR_026982.1 | 3 | 798 | ||||
GenBank mRNA | AK293095.1 | - | 3 | 521 | ||
BC065556.1 | - | 3 | 665 | |||
BQ668440.1 | - | 4 | 870 |
Gene Symbol: RPL23AP87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029406.1 | 4 | 1708 | |||
GenBank mRNA | AK125669.1 | - | 4 | 1706 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
