siRNA Details
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 20 | 2666 | ||
NM_173638.4 | NP_775909.2 | 17 | 2136 | |||
XM_017001077.1 | XP_016856566.1 | 15 | 1784 | |||
GenBank mRNA | AF379630.1 | - | 12 | 1314 | ||
AK097180.1 | - | 20 | 2585 | |||
AK290142.1 | - | 9 | 838 | |||
AK290302.1 | - | 14 | 1595 | |||
AK299360.1 | - | 11 | 1176 | |||
AY894566.1 | - | 14 | 1576 | |||
BC023087.1 | - | 17 | 1993 | 3918932 | ||
DA932476.1 | - | 4 | 270 |
Gene Symbol: NBPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094705.1 | - | 12 | 1344 |
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039703.5 | NP_001034792.4 | 2352 | |||
NM_001302371.1 | NP_001289300.1 | 2352 | ||||
GenBank mRNA | AF379632.1 | - | 7 | 610 | ||
AF380580.1 | - | 7 | 606 | |||
AF380582.1 | - | 3 | 244 | |||
AF419616.1 | - | 3 | 91 | |||
AK000726.1 | - | 18 | 2866 | |||
AL050141.1 | - | 1 | 820 | |||
AY894565.1 | - | 17 | 2164 | |||
BC021111.1 | - | 19 | 2396 | |||
BC063799.1 | - | 19 | 2379 | |||
BX538005.1 | - | 22 | 3092 |
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101663.4 | NP_001095133.3 | 20 | 2642 | ||
NM_183372.5 | NP_899228.4 | 22 | 3336 | |||
NR_046188.3 | 23 | 3404 | ||||
NR_110864.1 | 21 | 3328 | ||||
GenBank mRNA | AF419617.1 | - | 21 | 2755 | ||
AK054850.1 | - | 6 | 1081 | |||
AK302413.1 | - | 19 | 2478 | |||
AL832622.2 | - | 23 | 3539 | |||
BC136292.1 | - | 22 | 3336 | 9020279 | ||
BX648497.1 | - | 21 | 3127 |
Gene Symbol: NBPF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278141.1 | NP_001265070.1 | 29 | 4347 | ||
GenBank mRNA | AB033071.1 | - | 29 | 3820 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 18 | 2317 | ||
XM_011509395.2 | XP_011507697.1 | 21 | 2817 | |||
GenBank mRNA | AF161426.1 | - | 4 | 269 | ||
AK095459.1 | - | 4 | 1052 | |||
AL117237.1 | - | 12 | 1314 |
Gene Symbol: NBPF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003019.1 | XP_016858508.1 | 15 | 1777 | ||
GenBank mRNA | AF379633.1 | - | 3 | 177 | ||
AF379636.1 | - | 7 | 709 | |||
AK292666.1 | - | 6 | 530 | |||
CR936830.1 | - | 3 | 3190 |
Gene Symbol: NBPF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278267.1 | NP_001265196.1 | 1672 | |||
GenBank mRNA | AK299612.1 | - | 6 | 486 |
Gene Symbol: NBPF25P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104217.1 | 17 | 1961 | |||
GenBank mRNA | AK125792.1 | - | 17 | 1961 |
Gene Symbol: NBPF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542529.2 | XP_011540831.1 | 18 | 2347 | ||
XM_017003011.1 | XP_016858500.1 | 18 | 2346 | |||
GenBank mRNA | AK294944.1 | - | 21 | 2230 | ||
DA203219.1 | - | 5 | 340 | |||
DA675097.1 | - | 5 | 315 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037501.2 | NP_001032590.2 | 18 | 2489 | ||
NR_102404.1 | 17 | 2325 | ||||
NR_102405.1 | 16 | 2222 | ||||
GenBank mRNA | AY894561.1 | - | 11 | 1289 | ||
AY894562.1 | - | 13 | 1514 | |||
AY894563.1 | - | 12 | 1388 | |||
AY894567.1 | - | 16 | 2222 | |||
AY894569.1 | - | 17 | 2122 | |||
AY894570.1 | - | 13 | 1514 | |||
AY894571.1 | - | 17 | 2386 | |||
AY894572.1 | - | 17 | 2325 | |||
AY894573.1 | - | 17 | 2386 | |||
BC086308.1 | - | 18 | 2332 | 5453529 |
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037675.3 | NP_001032764.2 | 19 | 2392 | ||
NM_001277444.1 | NP_001264373.1 | 19 | 2392 | |||
XM_006711317.2 | XP_006711380.1 | 20 | 2620 | |||
GenBank mRNA | AF419618.1 | - | 12 | 1341 | ||
AK095030.1 | - | 5 | 3304 | |||
AK294414.1 | - | 13 | 1550 | |||
AY894574.1 | - | 19 | 2394 | |||
BC071995.1 | - | 14 | 1576 | |||
BC094841.1 | - | 14 | 1576 | |||
BQ436001.1 | - | 3 | 126 | |||
DQ786323.1 | - | 6 | 565 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
