siRNA Details
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008493.1 | NP_001008493.1 | 15 | 9657 | ||
NM_018212.4 | NP_060682.2 | 14 | 9594 | |||
XM_005273187.2 | XP_005273244.1 | 16 | 9697 | |||
XM_017001746.1 | XP_016857235.1 | 17 | 10932 | |||
XM_017001751.1 | XP_016857240.1 | 14 | 9942 | |||
XM_017001753.1 | XP_016857242.1 | 18 | 9736 | |||
XM_017001754.1 | XP_016857243.1 | 17 | 9680 | |||
XM_017001755.1 | XP_016857244.1 | 17 | 9423 | |||
XM_017001756.1 | XP_016857245.1 | 15 | 9354 | |||
XM_017001757.1 | XP_016857246.1 | 15 | 9257 |
Gene Symbol: ASB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123854.1 | - | 5 | 2174 |
Gene Symbol: BORCS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057161.1 | - | 1 | 1259 | ||
AK128256.1 | - | 1 | 1868 |
Gene Symbol: CARMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | GQ292874.1 | - | 1 | 539 |
Gene Symbol: CCPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC034914.1 | - | 1 | 521 | 5276323 |
Gene Symbol: CFHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK311681.1 | - | 2 | 362 |
Gene Symbol: CNTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006713004.3 | XP_006713067.1 | 24 | 5736 | ||
XM_011533425.2 | XP_011531727.1 | 24 | 5739 | |||
XM_011533426.2 | XP_011531728.1 | 25 | 5958 | |||
XM_011533427.2 | XP_011531729.1 | 25 | 5939 | |||
XM_011533428.2 | XP_011531730.1 | 23 | 5928 | |||
XM_011533429.2 | XP_011531731.1 | 23 | 5863 | |||
XM_011533430.2 | XP_011531732.1 | 23 | 5824 | |||
XM_011533431.2 | XP_011531733.1 | 22 | 8573 | |||
XM_017005782.1 | XP_016861271.1 | 25 | 5681 | |||
XM_017005783.1 | XP_016861272.1 | 26 | 6555 | |||
XM_017005784.1 | XP_016861273.1 | 24 | 6016 | |||
XM_017005785.1 | XP_016861274.1 | 25 | 5663 | |||
XM_017005786.1 | XP_016861275.1 | 23 | 5925 | |||
XM_017005787.1 | XP_016861276.1 | 23 | 5821 |
Gene Symbol: DAPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011521414.1 | XP_011519716.1 | 11 | 1261 | ||
XM_017022044.1 | XP_016877533.1 | 11 | 1257 | |||
GenBank mRNA | AK094290.1 | - | 1 | 319 | ||
BC114951.1 | - | 10 | 1876 | 40005391 |
Gene Symbol: DEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030567.1 | - | 3 | 414 |
Gene Symbol: DFFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282669.1 | NP_001269598.1 | 7 | 2499 | ||
NM_001320132.1 | NP_001307061.1 | 7 | 2596 | |||
NM_001320136.1 | NP_001307065.1 | 7 | 2477 | |||
NM_004402.3 | NP_004393.1 | 7 | 2427 | |||
NR_104222.1 | 8 | 2564 | ||||
NR_135150.1 | 8 | 2551 | ||||
NR_135151.1 | 8 | 2514 | ||||
NR_135152.1 | 7 | 2449 | ||||
XM_017000498.1 | XP_016855987.1 | 6 | 2362 | |||
XM_017000499.1 | XP_016855988.1 | 6 | 2378 | |||
XM_017000500.1 | XP_016855989.1 | 6 | 2133 | |||
GenBank mRNA | AB028911.1 | - | 8 | 2343 | ||
AB028912.1 | - | 7 | 2389 | |||
AB028913.1 | - | 8 | 2307 | |||
AF064019.1 | - | 7 | 2220 | |||
AK027141.1 | - | 2171 | ||||
BC032827.1 | - | 8 | 2376 | |||
BC048797.1 | - | 7 | 2413 | 5204996 |
Gene Symbol: DNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096629.1 | - | 1 | 65 |
Gene Symbol: KIAA1841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CA421892.1 | - | 1 | 429 |
Gene Symbol: LOC100131257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034022.1 | 1 | 14846 |
Gene Symbol: LOC101928780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128544.1 | - | 1 | 75 |
Gene Symbol: MTMR9LP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833686.1 | - | 7 | 2135 |
Gene Symbol: NANS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091184.1 | - | 1 | 1975 |
Gene Symbol: PASK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510834.1 | XP_011509136.1 | 1 | 22 | ||
GenBank mRNA | AL117400.1 | - | 1 | 1012 |
Gene Symbol: POLR1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK225050.1 | - | 13 | 1743 | ||
BC017766.1 | - | 3 | 712 |
Gene Symbol: PP12708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF318327.1 | - | 2 | 1851 |
Gene Symbol: SAMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091282.1 | - | 1 | 453 |
Gene Symbol: SAP30L-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DB100745.1 | - | 2 | 305 |
Gene Symbol: SGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024292.1 | - | 1 | 483 |
Gene Symbol: SHOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000451.3 | NP_000442.1 | 6 | 3075 | ||
GenBank mRNA | U89331.1 | - | 6 | 3075 |
Gene Symbol: SIRT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193267.2 | NP_001180196.1 | 9 | 1887 | ||
NM_001242827.1 | NP_001229756.1 | 9 | 1807 | |||
NM_012241.4 | NP_036373.1 | 10 | 1963 | |||
XM_005248968.4 | XP_005249025.1 | 10 | 1878 | |||
XM_011514444.2 | XP_011512746.1 | 11 | 2012 | |||
XM_017010621.1 | XP_016866110.1 | 9 | 1754 | |||
XM_017010622.1 | XP_016866111.1 | 11 | 2071 | |||
XM_017010623.1 | XP_016866112.1 | 11 | 2201 | |||
GenBank mRNA | AK094269.1 | - | 1 | 287 | ||
DR003695.1 | - | 1 | 425 |
Gene Symbol: SLC8B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095092.1 | - | 1 | 722 |
Gene Symbol: SYMPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209024.1 | - | 1 | 214 |
Gene Symbol: TATDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC048115.1 | - | 10 | 1439 |
Gene Symbol: TMEM266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308676.1 | - | 11 | 1640 |
Gene Symbol: WDR76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167941.1 | NP_001161413.1 | 13 | 2369 | ||
NM_024908.3 | NP_079184.2 | 13 | 2479 | |||
XM_005254680.3 | XP_005254737.1 | 13 | 2442 | |||
GenBank mRNA | AK023035.1 | - | 13 | 2384 | ||
BC051855.1 | - | 13 | 2383 | 6141090 | ||
EF011624.1 | - | 13 | 2383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
