siRNA Details
Gene Symbol: FAM163B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080515.2 | NP_001073984.1 | 2 | 388 | ||
XM_005272203.4 | XP_005272260.1 | 3 | 1265 | |||
XM_005272204.4 | XP_005272261.1 | 5 | 713 | |||
XM_006717233.3 | XP_006717296.1 | 3 | 675 | |||
XM_011518912.2 | XP_011517214.1 | 3 | 1209 | |||
XM_011518913.2 | XP_011517215.1 | 3 | 2298 | |||
XM_017015017.1 | XP_016870506.1 | 3 | 703 | |||
XM_017015018.1 | XP_016870507.1 | 3 | 971 | |||
GenBank mRNA | BC146946.1 | - | 2 | 388 | 9056874 | |
BC146961.1 | - | 2 | 388 | 9056889 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
