siRNA Details
Gene Symbol: HIST1H2BK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312653.1 | NP_001299582.1 | 1 | 247 | ||
NM_080593.2 | NP_542160.1 | 1 | 247 | |||
GenBank mRNA | AJ223352.1 | - | 1 | 203 | ||
AK292062.1 | - | 1 | 244 | |||
BC000893.1 | - | 1 | 229 | 3463930 | ||
BC051872.1 | - | 1 | 227 | 6526835 | ||
BC064959.1 | - | 1 | 227 | 6093977 | ||
BC108737.1 | - | 1 | 227 | 4707078 | ||
DA873786.1 | - | 1 | 247 |
Gene Symbol: H2BFS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC126339.1 | - | 1 | 237 | ||
BC126369.1 | - | 1 | 237 |
Gene Symbol: HIST1H2BL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003519.3 | NP_003510.1 | 1 | 212 | ||
GenBank mRNA | AK312114.1 | - | 1 | 251 | ||
BC093759.1 | - | 1 | 269 | |||
BC093761.1 | - | 1 | 268 |
Gene Symbol: LOC102724334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006723918.3 | XP_006723981.1 | 1 | 211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
