siRNA Details
Gene Symbol: HIST1H2BK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312653.1 | NP_001299582.1 | 1 | 77 | ||
NM_080593.2 | NP_542160.1 | 1 | 77 | |||
GenBank mRNA | AJ223352.1 | - | 1 | 33 | ||
AK292062.1 | - | 1 | 74 | |||
BC000893.1 | - | 1 | 59 | 3463930 | ||
BC051872.1 | - | 1 | 57 | 6526835 | ||
BC064959.1 | - | 1 | 57 | 6093977 | ||
BC108737.1 | - | 1 | 57 | 4707078 | ||
DA873786.1 | - | 1 | 77 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
