siRNA Details
Gene Symbol: NEU4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167599.2 | NP_001161071.1 | 4 | 1885 | ||
NM_001167600.2 | NP_001161072.1 | 4 | 1562 | |||
NM_001167601.2 | NP_001161073.1 | 4 | 1565 | |||
NM_001167602.2 | NP_001161074.1 | 5 | 2063 | |||
NM_080741.3 | NP_542779.2 | 4 | 1882 | |||
GenBank mRNA | AJ277883.1 | - | 3 | 1349 | ||
AK025617.1 | - | 5 | 1815 | |||
AK091038.1 | - | 5 | 2060 | |||
AK096992.1 | - | 2 | 2675 | |||
AK289421.1 | - | 4 | 1439 | |||
AK290076.1 | - | 4 | 1422 | |||
AK298403.1 | - | 5 | 1394 | |||
AK300969.1 | - | 4 | 1435 | |||
AY203952.1 | - | 4 | 1417 | |||
BC012899.1 | - | 4 | 1836 | 4156395 | ||
BC105974.1 | - | 4 | 1398 | 4944243 | ||
BC117214.1 | - | 4 | 1526 | 40125765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
